Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • S100B Antibodies

NeoBiotechnologies

S100B (Astrocyte and Melanoma Marker) Monoclonal Antibody (4C4.9, S100B/1012)

View all (97) S100B antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite S100B (Astrocyte and Melanoma Marker) Monoclonal Antibody (4C4.9, S100B/1012)

S100B (Astrocyte and Melanoma Marker) Monoclonal Antibody (4C4.9, S100B/1012)
  • Antibody Testing Data (3)
S100B (Astrocyte and Melanoma Marker) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
S100B (Astrocyte and Melanoma Marker) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

S100B (Astrocyte and Melanoma Marker) Antibody (6285-MSM3-P0) in ICC/IF

Confocal Immunofluorescent analysis of A2058 cells using AF488-labeled Isotype Control Monoclonal Antibody (IgG2a) (Green). F-actin filaments were labeled with DyLight 554 Phalloidin (red). DAPI was used to stain the cell nuclei (blue). (Negative Control). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

S100B (Astrocyte and Melanoma Marker) Antibody in Immunocytochemistry (ICC/IF)
S100B (Astrocyte and Melanoma Marker) Antibody in Immunocytochemistry (ICC/IF)
S100B (Astrocyte and Melanoma Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))

Product Details

6285-MSM3-P0

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

0.25-0.5 µg/mL
-

Immunocytochemistry (ICC/IF)

1-2 µg/mL
-

Flow Cytometry (Flow)

1-2 µg/10^6 cells
-

Immunomicroscopy (IM)

Assay-dependent
-
Product Specifications

Species Reactivity

Bovine, Human, Mouse, Rat

Host/Isotype

Mouse / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

4C4.9, S100B/1012

Immunogen

Purified bovine brain S100 protein (4C4.9). Recombinant full-length human S100B protein (S100B/1012)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Wet ice

Target Information

S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer's disease, Down's syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Protein S100-B; S-100 calcium-binding protein, beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S100 protein beta chain; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural

View more View less

Gene Aliases: AI850290; BOS_1447; Bpb; NEF; S100; S100-B; S100B; S100beta; S100P

View more View less

UniProt ID: (Human) P04271, (Bovine) P02638, (Mouse) P50114, (Rat) P04631

View more View less

Entrez Gene ID: (Human) 6285, (Bovine) 525716, (Mouse) 20203, (Rat) 25742

View more View less

Function(s)
calcium ion binding protein binding zinc ion binding identical protein binding protein homodimerization activity S100 protein binding tau protein binding calcium-dependent protein binding RAGE receptor binding kinase inhibitor activity receptor binding metal ion binding
Process(es)
axonogenesis central nervous system development learning or memory memory cell proliferation positive regulation of cell proliferation regulation of cell shape positive regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling innate immune response regulation of neuronal synaptic plasticity astrocyte differentiation response to glucocorticoid response to methylmercury long-term synaptic potentiation cellular response to hypoxia negative regulation of skeletal muscle cell differentiation regulation of translation phosphorylation negative regulation of phosphorylation positive regulation of complement activation astrocyte activation energy reserve metabolic process cellular calcium ion homeostasis regulation of cytokine biosynthetic process regulation of long-term neuronal synaptic plasticity positive regulation of synaptic transmission
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5f5c96ff6c-st5hm:80/100.66.129.203:80.
git-commit: 945c3ce0b94337c0c8e9574aed9898c23d0d1109
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0