Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • RPA2 Antibodies

OriGene

RPA2 Monoclonal Antibody (OTI8B3), TrueMAB™

View all (63) RPA2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite RPA2 Monoclonal Antibody (OTI8B3), TrueMAB™

  • Antibody Testing Data (15)
RPA2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
RPA2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 15

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

RPA2 Antibody (TA500788) in ICC/IF

Anti-RPA2 mouse monoclonal antibody (TA500788) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY RPA2(RC205715). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

RPA2 Antibody in Immunocytochemistry (ICC/IF)
RPA2 Antibody in Immunocytochemistry (ICC/IF)
RPA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RPA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RPA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RPA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RPA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RPA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RPA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RPA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RPA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RPA2 Antibody in Western Blot (WB)
RPA2 Antibody in Flow Cytometry (Flow)
RPA2 Antibody in Flow Cytometry (Flow)
RPA2 Antibody in Immunoprecipitation (IP)
RPA2 Monoclonal Antibody (OTI8B3), TrueMAB™

Product Details

TA500788

Applications
Tested Dilution
Publications

Western Blot (WB)

1:4,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50
-

Immunocytochemistry (ICC/IF)

1:50-1:100
-

Flow Cytometry (Flow)

1:100
-

Immunoprecipitation (IP)

2 µg/500 µL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI8B3

Immunogen

Full length human recombinant protein of human RPA2 produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.6 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: HSSB; p32; p34; Replication factor A protein 2; Replication protein A 32 kDa subunit; Replication protein A 32kDa subunit; Replication protein A 34 kDa subunit; Replication protein A 34kDa subunit; replication protein A2, 32kDa; RF-A protein 2; RFA; RP-A p32; RP-A p34; RP21C; RP4-547C9.3; RpLP1; RpP2

View more View less

Gene Aliases: REPA2; RP-A p32; RP-A p34; RPA2; RPA32; RPA34

View more View less

UniProt ID: (Human) P15927

View more View less

Entrez Gene ID: (Human) 6118

View more View less

Function(s)
damaged DNA binding single-stranded DNA binding protein binding enzyme binding protein phosphatase binding ubiquitin protein ligase binding protein N-terminus binding
Process(es)
G1/S transition of mitotic cell cycle telomere maintenance via recombination telomere maintenance double-strand break repair via homologous recombination DNA replication transcription-coupled nucleotide-excision repair base-excision repair nucleotide-excision repair nucleotide-excision repair, preincision complex stabilization nucleotide-excision repair, preincision complex assembly nucleotide-excision repair, DNA incision, 3'-to lesion nucleotide-excision repair, DNA incision, 5'-to lesion nucleotide-excision repair, DNA gap filling mismatch repair regulation of double-strand break repair via homologous recombination translesion synthesis mitotic G1 DNA damage checkpoint nucleotide-excision repair, DNA incision interstrand cross-link repair error-prone translesion synthesis DNA damage response, detection of DNA damage error-free translesion synthesis regulation of cellular response to heat regulation of signal transduction by p53 class mediator regulation of DNA damage checkpoint
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0