Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • RNH1 Antibodies

Invitrogen

RNH1 Polyclonal Antibody

View all (26) RNH1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite RNH1 Polyclonal Antibody

  • Antibody Testing Data (10)
RNH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
RNH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 10

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

RNH1 Antibody (PA5-79931) in IHC (P)

Immunohistochemical analysis of RNH1 in paraffin-embedded section of human intestinal cancer tissues. Heat mediated antigen retrieval was performed in citrate buffer (pH6, epitope retrieval solution) for 20 mins. The tissue section was blocked with 10% goat serum. The tissue section was then incubated with 1μg/mL rabbit anti-RNH1 antibody (Product # PA5-79931) overnight at 4°C. Biotinylated goat anti-rabbit... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

RNH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RNH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RNH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RNH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
RNH1 Antibody in Western Blot (WB)
RNH1 Antibody in Western Blot (WB)
RNH1 Antibody in Flow Cytometry (Flow)
RNH1 Antibody in Flow Cytometry (Flow)
RNH1 Antibody in Flow Cytometry (Flow)
RNH1 Antibody in Flow Cytometry (Flow)
RNH1 Polyclonal Antibody

Product Details

PA5-79931

Applications
Tested Dilution
Publications

Western Blot (WB)

0.1-0.5 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

0.5-1 µg/mL
-

Immunohistochemistry (Frozen) (IHC (F))

0.5-1 µg/mL
-

Immunocytochemistry (ICC/IF)

0.5-1 µg/mL
-

Flow Cytometry (Flow)

1-3 µg/1x10^6 cells
-
Product Specifications

Species Reactivity

Human, Mouse, Non-human primate, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

A synthetic peptide corresponding to a sequence at the N-terminus of human RNH1 (2-37aa SLDIQSLDIQCEELSDARWAELLPLLQQCQVVRLDD).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

500 µg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS with 5mg BSA

Contains

0.05mg sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2747046

Product Specific Information

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

Positive Control - WB: human placenta tissue, human Hela whole cell, human SK-OV-3 whole cell, human Jurkat whole cell, human U-87MG whole cell, monkey COS-7 whole cell, human SW620 whole cell, human Caco-2 whole cell, rat brain tissue, rat testis tissue, rat lung tissue, mouse brain tissue, mouse testis tissue, mouse lung tissue, mouse HEPA1-6 whole cell. IHC: human intestinal cancer tissue, human intestinal cancer tissue, human lung cancer tissue, human lung cancer tissue. Flow: A549 cell, A549 cell.

Target Information

RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Placental ribonuclease inhibitor; Placental RNase inhibitor; RAI; Ribonuclease inhibitor; Ribonuclease/angiogenin inhibitor 1; testicular tissue protein Li 164

View more View less

Gene Aliases: AW546468; C80305; PRI; RAI; RNH; RNH1

View more View less

UniProt ID: (Human) P13489, (Mouse) Q91VI7, (Rat) P29315

View more View less

Entrez Gene ID: (Human) 6050, (Mouse) 107702, (Rat) 100360501

View more View less

Function(s)
protein binding ribonuclease inhibitor activity
Process(es)
mRNA catabolic process negative regulation of catalytic activity regulation of angiogenesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0