Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Protein C Antibodies

Invitrogen

Protein C Recombinant Mouse Monoclonal Antibody (HPC-4)

View all (25) Protein C antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Protein C Recombinant Mouse Monoclonal Antibody (HPC-4)

  • Antibody Testing Data (3)
Protein C Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Protein C Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Protein C Antibody (MA5-51846) in ICC/IF

Immunocytochemistry analysis of Protein C in paraformaldehyde fixed HEK293 cells transfected with Protein C expressing plasmid (A) and non-transfected HEK293 cells (B), permeabilized with 0.15% Triton using Protein C Recombinant Mouse Monoclocal Antibody (Product # MA5-51846) at a 1:200 dilution for 1h followed by Alexa Fluor 488 secondary antibody (1:1,000 dilution), showing membrane and cytoplasmic staini... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Protein C Antibody in Immunocytochemistry (ICC/IF)
Protein C Antibody in Western Blot (WB)
Protein C Antibody in ELISA (ELISA)

Product Details

MA5-51846

Applications
Tested Dilution
Publications

Western Blot (WB)

Assay-dependent
-

Immunocytochemistry (ICC/IF)

1:200
-

ELISA (ELISA)

10 µg/mL
-

Inhibition Assays (IA)

Assay-dependent
-

Affinity Purification (AP)

Assay-dependent
-
Product Specifications

Species Reactivity

Baboon, Human

Host/Isotype

Mouse / IgG1, kappa

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

HPC-4

Immunogen

This antibody was raised by immunising mice with human protein C.

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS

Contains

0.02% ProClin 300

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3250538

Product Specific Information

Specifically binds to a specific twelve peptide sequence (EDQVDPRLIDGK) in the activation region of the Protein C (spanning the thrombin cleavage site). Does not react with activated protein C. Has been shown to be cross-reactive with baboon Protein C but not with bovine or mouse Protein C.

Target Information

Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55 kDa and the normal concentrations in human plasma is approximately 1-3 ng/mL because of the very fast turnover, the proenzyme concentration is approximately 3 µg/mL. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulminans, and recurrent venous thrombosis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Anticoagulant protein C; Autoprothrombin IIA; Blood coagulation factor XIV; EDQVDPRLIDGK; prepro-protein C; protein C (inactivator of coagulation factors Va and VIIIa); type I protein C; Vitamin K-dependent protein C

View more View less

Gene Aliases: APC; PC; PROC; PROC1; THPH3; THPH4

View more View less

UniProt ID: (Human) P04070

View more View less

Entrez Gene ID: (Human) 5624

View more View less

Function(s)
serine-type endopeptidase activity calcium ion binding protein binding
Process(es)
signal peptide processing proteolysis ER to Golgi vesicle-mediated transport blood coagulation peptidyl-glutamic acid carboxylation negative regulation of blood coagulation negative regulation of apoptotic process negative regulation of inflammatory response negative regulation of coagulation leukocyte migration positive regulation of establishment of endothelial barrier
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0