Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • NMDAR1 Antibodies

Invitrogen

Phospho-NMDAR1 (Ser890) Polyclonal Antibody

View all (38) NMDAR1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-NMDAR1 (Ser890) Polyclonal Antibody

  • Antibody Testing Data (3)
Phospho-NMDAR1 (Ser890) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Phospho-NMDAR1 (Ser890) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-NMDAR1 (Ser890) Antibody (PA5-104969) in ICC/IF

Immunofluorescent analysis of Phospho-NMDAR1 (Ser890) in A549 cells. Samples were fixed with paraformaldehyde, permeabilized with 0.1% saponin, blocked with 10% serum (45 min, 37°C) incubated with Phospho-NMDAR1 (Ser890) polyclonal antibody (Product # PA5-104969) using a dilution of 1:400 (1 hr, 37°C), and followed by goat anti-rabbit IgG Alexa Fluor 594 at a dilution of 1:600. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-NMDAR1 (Ser890) Antibody in Immunocytochemistry (ICC/IF)
Phospho-NMDAR1 (Ser890) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Phospho-NMDAR1 (Ser890) Antibody in Western Blot (WB)
Phospho-NMDAR1 (Ser890) Polyclonal Antibody

Product Details

PA5-104969

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunocytochemistry (ICC/IF)

1:100-1:500
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

A synthesized peptide derived from human GRIN1(Accession Q05586), corresponding to amino acid residues around phosphorylated Ser890.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

sequential chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

RRID

AB_2816442

Product Specific Information

Antibody detects endogenous levels of NMDAR1 only when phosphorylated at Serine 890.

Target Information

NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: GluN1; Glun1r; glutamate [NMDA] receptor subunit zeta 1; Glutamate [NMDA] receptor subunit zeta-1; glutamate receptor; Glutamate receptor ionotropic, NMDA 1; glutamate receptor, ionotropic, N-methyl D-aspartate 1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; neurotransmitter receptor; NMD-R1; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDZ1; NMZ1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; RP11-350O14.1

View more View less

Gene Aliases: GluN1; GluRdelta1; Glurz1; GluRzeta1; GRIN1; M100174; MRD8; NMD-R1; NMDA1; Nmdar; NMDAR1; NR1; Rgsc174

View more View less

UniProt ID: (Human) Q05586, (Rat) P35439, (Mouse) P35438

View more View less

Entrez Gene ID: (Human) 2902, (Rat) 24408, (Mouse) 14810

View more View less

Function(s)
NMDA glutamate receptor activity Ras guanyl-nucleotide exchange factor activity extracellular-glutamate-gated ion channel activity calcium channel activity calcium ion binding protein binding calmodulin binding glycine binding glutamate binding enzyme binding voltage-gated cation channel activity glutamate receptor binding neurotransmitter binding protein heterodimerization activity ionotropic glutamate receptor activity receptor binding cation channel activity protein dimerization activity receptor activity ion channel activity
Process(es)
MAPK cascade conditioned taste aversion suckling behavior response to amphetamine cation transport cellular calcium ion homeostasis chemical synaptic transmission respiratory gaseous exchange long-term memory adult locomotory behavior olfactory learning visual learning propylene metabolic process sensory perception of pain pons maturation cerebral cortex development regulation of ion transmembrane transport social behavior ionotropic glutamate receptor signaling pathway synaptic transmission, glutamatergic regulation of membrane potential positive regulation of apoptotic process response to morphine negative regulation of neuron apoptotic process positive regulation of GTPase activity regulation of respiratory gaseous exchange response to ethanol positive regulation of transcription from RNA polymerase II promoter ephrin receptor signaling pathway regulation of long-term neuronal synaptic plasticity rhythmic process regulation of dendrite morphogenesis regulation of axonogenesis protein tetramerization response to calcium ion regulation of synapse assembly calcium ion homeostasis excitatory postsynaptic potential prepulse inhibition male mating behavior response to fungicide calcium ion transmembrane transport cellular response to manganese ion positive regulation of excitatory postsynaptic potential startle response calcium ion transport learning or memory learning memory associative learning regulation of cell communication positive regulation of cell death response to organic cyclic compound response to amine ion transmembrane transport regulation of neuron apoptotic process regulation of synaptic plasticity regulation of neuronal synaptic plasticity neuromuscular process transport ion transport cation transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0