Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ERK1/2 Antibodies

Invitrogen

Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Recombinant Rabbit Monoclonal Antibody (SC58-01)

Advanced Verification
View all (85) ERK1/2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Recombinant Rabbit Monoclonal Antibody (SC58-01)

  • Antibody Testing Data (10)
  • Advanced Verification (3)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 13

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody (MA5-55603) in ICC/IF

Immunofluorescence analysis of Phospho-Erk1 (T202+Y204)+Erk2 (T185+Y187) using NIH/3T3 cells treated with or without 200 nM PMA for 30 minutes. The cells were fixed 4% paraformaldehyde for 20 minutes, permeabilized with 0.1% Triton™ X-100 in PBS for 5 minutes, and blocked with 1% BSA in 10% negative goat serum for 1 hour at room temperature. The cells were labeled with Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Immunocytochemistry (ICC/IF)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Immunocytochemistry (ICC/IF)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Western Blot (WB)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Western Blot (WB)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Western Blot (WB)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Western Blot (WB)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Western Blot (WB)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Western Blot (WB)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody in Flow Cytometry (Flow)
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Antibody
Phospho-ERK1/ERK2 (Thr202, Tyr204, Thr185, Tyr187) Recombinant Rabbit Monoclonal Antibody (SC58-01)

Product Details

MA5-55603

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:1,000
-

Immunocytochemistry (ICC/IF)

1:100
-

Flow Cytometry (Flow)

1:1,000
-

Immunoprecipitation (IP)

Assay-dependent
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

SC58-01

Immunogen

Synthetic peptide within human aa 166-215/379.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

TBS, pH 7.4, with 0.05% BSA, 40% glycerol

Contains

0.05% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Product Specific Information

Positive Control: SH-SY5Y cell lysate, SH-SY5Y treated with 100 ng/mL hBeta-NGF for 10 minutes cell lysate, PC-12 treated with 100 ng/mL hBeta-NGF for 10 minutes cell lysate, SK-Br-3 whole cell lysate, NIH/3T3 cell lysate, NIH/3T3 treated with 200 nM PMA for 30 minutes cell lysate, HeLa treated with 200 nM PMA for 30 minutes cell lysate, human thyroid carcinoma tissue.

Tissue Specificity: Low tissue specificity.

Subcellular Location: Cytoplasm, Nucleus.

Sequence Similarities: 100% Mouse/Rat.

Predicted band size: 41/43 kDa.

Target Information

ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: ERK-1; ERK-2; erk1 erk2; ERK1b; ERT1; ERT2; Extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-related kinase 1; extracellular-signal-regulated kinase 2; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MAPK 1; MAPK 2; MAPK 3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase 1; mitogen-activated 3; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; MNK1; p42; p42 MAP Kinase; p42-MAPK; p44 MAP kinase; p44-ERK1; p44-MAPK; pp42/MAP kinase; protein tyrosine kinase ERK2

View more View less

Gene Aliases: 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK-1; ERK-2; ERK1; ERK2; ERT1; ERT2; Esrk1; HS44KDAP; HUMKER1A; Mapk; MAPK1; MAPK2; MAPK3; MNK1; Mtap2k; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; p44; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; PRKM1; PRKM2; PRKM3

View more View less

UniProt ID: (Human) P28482, (Human) P27361, (Mouse) P63085, (Rat) P63086, (Rat) P21708, (Mouse) Q63844

View more View less

Entrez Gene ID: (Human) 5594, (Human) 5595, (Mouse) 26413, (Rat) 116590, (Rat) 50689, (Mouse) 26417

View more View less

Function(s)
phosphotyrosine binding DNA binding protein serine/threonine kinase activity MAP kinase activity protein binding ATP binding transcription factor binding RNA polymerase II carboxy-terminal domain kinase activity kinase activity phosphatase binding mitogen-activated protein kinase kinase kinase binding identical protein binding scaffold protein binding nucleotide binding protein kinase activity transferase activity transferase activity, transferring phosphorus-containing groups protein kinase binding non-receptor serine/threonine protein kinase
Process(es)
MAPK cascade activation of MAPK activity MAPK import into nucleus transcription, DNA-templated protein phosphorylation apoptotic process chemotaxis response to stress cellular response to DNA damage stimulus cell cycle signal transduction chemical synaptic transmission axon guidance positive regulation of cell proliferation fibroblast growth factor receptor signaling pathway response to toxic substance positive regulation of peptidyl-threonine phosphorylation regulation of phosphatidylinositol 3-kinase signaling viral process peptidyl-serine phosphorylation peptidyl-threonine phosphorylation sensory perception of pain cytosine metabolic process platelet activation positive regulation of cell migration thyroid gland development regulation of protein stability lipopolysaccharide-mediated signaling pathway positive regulation of telomere maintenance via telomerase regulation of stress-activated MAPK cascade mammary gland epithelial cell proliferation Fc-epsilon receptor signaling pathway Fc-gamma receptor signaling pathway involved in phagocytosis ERBB signaling pathway outer ear morphogenesis response to exogenous dsRNA response to estrogen negative regulation of cell differentiation positive regulation of translation positive regulation of transcription, DNA-templated thymus development T cell receptor signaling pathway B cell receptor signaling pathway regulation of sequence-specific DNA binding transcription factor activity regulation of cytoskeleton organization positive regulation of telomerase activity Bergmann glial cell differentiation long-term synaptic potentiation face development JAK-STAT cascade involved in growth hormone signaling pathway lung morphogenesis trachea formation labyrinthine layer blood vessel development cardiac neural crest cell development involved in heart development ERK1 and ERK2 cascade response to epidermal growth factor caveolin-mediated endocytosis regulation of Golgi inheritance cellular response to granulocyte macrophage colony-stimulating factor stimulus regulation of cellular response to heat positive regulation of telomere capping regulation of early endosome to late endosome transport positive regulation of protein phosphorylation transcription initiation from RNA polymerase I promoter protein complex assembly DNA damage induced protein phosphorylation phosphorylation arachidonic acid metabolic process BMP signaling pathway positive regulation of cyclase activity positive regulation of histone phosphorylation positive regulation of histone acetylation peptidyl-tyrosine autophosphorylation positive regulation of transcription from RNA polymerase II promoter cartilage development positive regulation of ERK1 and ERK2 cascade interleukin-1-mediated signaling pathway cellular response to mechanical stimulus positive regulation of xenophagy negative regulation of apolipoprotein binding organ morphogenesis response to lipopolysaccharide intracellular signal transduction cellular response to organic substance
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0