Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cyclin B3 Antibodies

Invitrogen

Phospho-Cyclin B3 (Ser284) Polyclonal Antibody

View all (13) Cyclin B3 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-Cyclin B3 (Ser284) Polyclonal Antibody

  • Antibody Testing Data (1)
Phospho-Cyclin B3 (Ser284) Antibody in Dot Blot (DB)
Group 53 Created with Sketch.
Phospho-Cyclin B3 (Ser284) Antibody in Dot Blot (DB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-Cyclin B3 (Ser284) Antibody (PA5-72387) in DB

Dot blot analysis of Phospho-Cyclin B3 (Ser284). Samples were incubated with Phospho-Cyclin B3 (Ser284) polyclonal antibody (Product # PA5-72387) using a dilution of 0.6 µg/mL. 50 ng of Phospho-peptide or Non Phospho-peptide per dot were adsorbed on nitrocellulose membrane. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-Cyclin B3 (Ser284) Antibody in Dot Blot (DB)

Product Details

PA5-72387

Applications
Tested Dilution
Publications

Dot blot (DB)

1:1,000
-
Product Specifications

Species Reactivity

Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S284 of rat CCNB3

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.3 mg/mL

Purification

Protein A, Antigen affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

0.09% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2718241

Target Information

Cell proliferation is controlled at specific stages of the cell cycle by distinct protein kinase complexes. These complexes consist of a catalytic subunit associating with a specific regulatory subunit to form the active kinase. The cyclins, which include cyclin A, B, C, D, E, F, G, H, I, K, L, T and their related proteins, including Dbf4, comprise the regulatory subunits of these kinase complexes. The controlled activation of the kinase complexes at various intervals of the cell cycle is regulated by the availability of the cyclins to the catalytic subunit. Unlike the catalytic subunit, which is expressed continually, the expression and stability of the regulatory subunit fluctuates depending on the stage of the cell cycle, thereby regulating kinase activity. Cyclin B3, also known as CCNB3 or CYCB3, is a 1,395 amino acid nuclear protein that belongs to the cyclin family of regulatory proteins. Expressed in testis with lower expression in a variety of other tissues, cyclin B3 is thought to be required for early meiotic prophase I, playing an important role in the meiotic cell cycle. Three isoforms of cyclin B3 exist due to alternative splicing events.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: cyclin B3; CyclinB3; G2/mitotic-specific cyclin-B3; Protein Ccnb3

View more View less

Gene Aliases: RGD1564367

View more View less

Entrez Gene ID: (Rat) 317389

View more View less

Function(s)
protein kinase binding
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity regulation of G2/M transition of mitotic cell cycle
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0