Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CrkL Antibodies

Invitrogen

Phospho-CrkL (Tyr207) Recombinant Rabbit Monoclonal Antibody (CrkLY207-G4), PE

View all (29) CrkL antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-CrkL (Tyr207) Recombinant Rabbit Monoclonal Antibody (CrkLY207-G4), PE

  • Antibody Testing Data (1)
Phospho-CrkL (Tyr207) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
Phospho-CrkL (Tyr207) Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-CrkL (Tyr207) Antibody (MA5-36970) in Flow

Flow cytometry of Phospho-CrkL (Tyr207) using K562 cells. Samples were incubated in monoclonal Phospho-CrkL (Tyr207) antibody (Product # MA5-36970). Images correspond with the following conditions: negative control (blue), imatinib (red), treated with pervanadate (green). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-CrkL (Tyr207) Antibody in Flow Cytometry (Flow)
Phospho-CrkL (Tyr207) Recombinant Rabbit Monoclonal Antibody (CrkLY207-G4), PE

Product Details

MA5-36970

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

5 µL/1x10^6 cells
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG, kappa

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

CrkLY207-G4

Immunogen

A synthetic phospho-peptide corresponding to residues surrounding Tyr207 of human phospho CrkL.
View immunogen

Conjugate

PE PE PE
  • Unconjugated
  • APC 
  • FITC
  • Request custom conjugation

Excitation/Emission Max

565/576 nm View spectra spectra

Form

Liquid

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.2% BSA

Contains

0.09% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2896905

Product Specific Information

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Target Information

The CRKL gene encodes the CRK-like protein, an adaptor protein that plays a crucial role in intracellular signaling pathways related to cell proliferation, differentiation, and migration. CRKL contains SH2 and SH3 domains that facilitate interactions with other signaling molecules, influencing multiple pathways such as the MAPK, PI3K, and STAT pathways. It is involved in processes like cytoskeleton reorganization and immune cell signaling, with significant implications for development and cancer. CRKL is notably associated with Philadelphia chromosome-positive leukemias, where it interacts with the BCR-ABL fusion protein, contributing to leukemogenesis. Additionally, dysregulation of CRKL can affect cardiovascular development, leading to congenital heart defects. Research into CRKL focuses on understanding its role in signal transduction and its potential as a therapeutic target due to its involvement in oncogenic and developmental pathways.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Crk-like protein; OTTHUMP00000198539; v-crk avian sarcoma virus CT10 oncogene homolog-like

View more View less

Gene Aliases: 1110025F07Rik; AA589403; AI325100; CRKL; Crkol

View more View less

UniProt ID: (Human) P46109, (Mouse) P47941

View more View less

Entrez Gene ID: (Human) 1399, (Mouse) 12929

View more View less

Function(s)
signal transducer activity SH3/SH2 adaptor activity protein binding poly(A) RNA binding cadherin binding involved in cell-cell adhesion
Process(es)
blood vessel development pattern specification process heart development organ morphogenesis anterior/posterior pattern specification thymus development parathyroid gland development activation of MAPKK activity JNK cascade Ras protein signal transduction positive regulation of cell proliferation intracellular signal transduction cell-cell adhesion positive regulation of substrate adhesion-dependent cell spreading
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0