Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • C/EBP beta Antibodies

Invitrogen

Phospho-C/EBP beta (Thr235) Polyclonal Antibody

View all (24) C/EBP beta antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-C/EBP beta (Thr235) Polyclonal Antibody

  • Antibody Testing Data (1)
Phospho-C/EBP beta (Thr235) Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Phospho-C/EBP beta (Thr235) Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-C/EBP beta (Thr235) Antibody (PA5-117238) in WB

Western blot analysis of Phospho-C/EBP beta (Thr235) in extracts of NIH/3T3 cells. Samples were incubated with Phospho-C/EBP beta (Thr235) Polyclonal antibody (Product # PA5-117238) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) secondary antibody at a dilution of 1:10,000. NIH/3T3 cells were treated by CIP (20 µL/400 µL) at 37°C for 1 hour. NIH/3T3 cells were treated by Serum-starv... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-C/EBP beta (Thr235) Antibody in Western Blot (WB)
Phospho-C/EBP beta (Thr235) Polyclonal Antibody

Product Details

PA5-117238

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

A synthetic phosphorylated peptide around T235 of human C/EBPB (NP_0051852)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.37 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2901868

Product Specific Information

Positive Samples: NIH/3T3

Immunogen sequence: PGTPS

Target Information

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: AGP/EBP; C EBP; C/EBP BETA; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer-binding protein beta; IL-6DBP; interleukin 6-dependent DNA-binding protein; Interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear protein Il6; OTTMUSP00000017503; TCF-5; Transcription factor 5; transcription factor C/EBP beta

View more View less

Gene Aliases: C/EBP-beta; C/EBPbeta; CEBPB; CRP2; IL-6DBP; IL6DBP; LAP; LIP; NF-IL6; NF-M; Nfil6; PP9092; TCF5

View more View less

UniProt ID: (Human) P17676, (Mouse) P28033

View more View less

Entrez Gene ID: (Human) 1051, (Mouse) 12608

View more View less

Function(s)
RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding protein binding transcription factor binding kinase binding histone acetyltransferase binding glucocorticoid receptor binding protein homodimerization activity histone deacetylase binding ubiquitin-like protein ligase binding protein heterodimerization activity sequence-specific DNA binding
Process(es)
ovarian follicle development embryonic placenta development granuloma formation transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter cell differentiation neuron differentiation response to lipopolysaccharide positive regulation of interleukin-4 production mammary gland epithelial cell proliferation response to endoplasmic reticulum stress T-helper 1 cell activation negative regulation of T cell proliferation defense response to bacterium negative regulation of neuron apoptotic process regulation of interleukin-6 biosynthetic process fat cell differentiation positive regulation of fat cell differentiation positive regulation of osteoblast differentiation regulation of osteoclast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter brown fat cell differentiation mammary gland epithelial cell differentiation regulation of transcription involved in cell fate commitment intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress cellular response to amino acid stimulus hepatocyte proliferation liver regeneration positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress acute-phase response inflammatory response immune response memory cellular response to lipopolysaccharide cellular response to interleukin-1 cellular response to organic cyclic compound
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0