Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Pannexin 1 Antibodies

Invitrogen

Pannexin 1 Recombinant Rabbit Monoclonal Antibody (28G5)

View all (14) Pannexin 1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Pannexin 1 Recombinant Rabbit Monoclonal Antibody (28G5)

  • Antibody Testing Data (4)
Pannexin 1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Pannexin 1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Pannexin 1 Antibody (MA5-50140) in ICC/IF

Immunocytochemistry analysis of U251 with Pannexin 1 monoclonal antibody (Product # MA5-50140) at 1:10, counter-stained with DAPI. The cells were fixed in 4% formaldehyde and blocked in 10% normal Goat Serum. The cells were then incubated with the antibody overnight at 4°C. The secondary antibody was Alexa Fluor 502-congugated AffiniPure Goat Anti-Rabbit IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Pannexin 1 Antibody in Immunocytochemistry (ICC/IF)
Pannexin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Pannexin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Pannexin 1 Antibody in Flow Cytometry (Flow)

Product Details

MA5-50140

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

Flow Cytometry (Flow)

1:50-1:200
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

28G5

Immunogen

A synthesized peptide derived from Human PANX1

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.21 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3092513

Target Information

The pannexin gene family encodes a second class of putative gap junction proteins and are highly conserved in invertebrates and mammals. Pannexins (Panx) are four-pass transmembrane proteins that oligomerize to form large pore ion and metabolite-permeable channels. Pannexin-1 (PANX1) and Pannexin-3 are closely related, while Pannexin-2 is a more distant relation. PANX1 is a transmembrane protein that forms a mechanosensitive ATP-permeable channel between adjacent cells and in the endoplasmic reticulum. PANX1 may play a role as a Ca2+ -leak channel to regulate ER Ca2+ homeostasis and regulates neural stem and progenitor cell proliferation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: innexin; Pannexin-1; PANX1

View more View less

Gene Aliases: MRS1; PANX1; PX1; UNQ2529; UNQ2529/PRO6028

View more View less

UniProt ID: (Human) Q543A0

View more View less

Entrez Gene ID: (Human) 24145

View more View less

Function(s)
protease binding receptor binding calcium channel activity leak channel activity ion channel binding protein heterodimerization activity actin filament binding gap junction hemi-channel activity scaffold protein binding
Process(es)
response to ischemia cation transport calcium ion transport cell-cell signaling response to ATP protein hexamerization positive regulation of interleukin-1 alpha secretion positive regulation of interleukin-1 beta secretion calcium ion transmembrane transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0