Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • FAPP2 Antibodies

Abnova

PLEKHA8 Polyclonal Antibody, MaxPab™

View all (6) FAPP2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PLEKHA8 Polyclonal Antibody, MaxPab™

  • Antibody Testing Data (1)
PLEKHA8 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
PLEKHA8 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PLEKHA8 Antibody (H00084725-D01P) in WB

Western Blot analysis of PLEKHA8 expression in transfected 293T cell line by PLEKHA8 MaxPab polyclonal antibody. Lane 1: PLEKHA8 transfected lysate(49.50 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PLEKHA8 Antibody in Western Blot (WB)
PLEKHA8 Polyclonal Antibody, MaxPab™

Product Details

H00084725-D01P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

PLEKHA8 (NP_116028.1, 1 a.a. approximately 440 a.a) full-length human protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Product may be used with Western Blot (Transfected lysate).

The Isotype of this product is composed of an IgG Mixture.

Immunogen sequence: MEGVLYKWTN YLSGWQPRWF LLCGGILSYY DSPEDAWKGC KGSIQMAVCE IQVHSVDNTR MDLIIPGEQY FYLKARSVAE RQRWLVALGS AKACLTDSRT QKEKEFAENT ENLKTKMSEL RLYCDLLVQQ VDKTKEVTTT GVSNSEEGID VGTLLKSTCN TFLKTLEECM QIANAAFTSE LLYRTPPGSP QLAMLKSSKM KHPIIPIHNS LERQMELSTC ENGSLNMEIN GEEEILMKNK NSLYLKSAEI DCSISSEENT DDNITVQGEI RKEDGMENLK NHDNNLTQSG SDSSCSPECL WEEGKEVIPT FFSTMNTSFS DIELLEDSGI PTEAFLASCY AVVPVLDKLG PTVFAPVKMD LVGNIKKVNQ KYITNKEEFT TLQKIVLHEV EADVAQVRNS ATEALLWLKR GLKFLKGFLT EVKNGEKDIQ TALRNPTENT

Target Information

Cargo transport protein that is required for apical transport from the Golgi complex. Transports AQP2 from the trans-Golgi network (TGN) to sites of AQP2 phosphorylation. Mediates the non-vesicular transport of glucosylceramide (GlcCer) from the trans-Golgi network (TGN) to the plasma membrane and plays a pivotal role in the synthesis of complex glycosphingolipids. Binding of both phosphatidylinositol 4-phosphate (PIP) and ARF1 are essential for the GlcCer transfer ability. Also required for primary cilium formation, possibly by being involved in the transport of raft lipids to the apical membrane, and for membrane tubulation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: FAPP-2; hFAPP2; MGC3358; PH domain-containing family A member 8; Phosphatidylinositol-four-phosphate adapter protein 2; phosphoinositol 4-phosphate adapter protein 2; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8; Pleckstrin homology domain-containing family A member 8; Serologically defined breast cancer antigen NY-BR-86

View more View less

Gene Aliases: FAPP2; PLEKHA8

View more View less

UniProt ID: (Human) Q96JA3

View more View less

Entrez Gene ID: (Human) 84725

View more View less

Function(s)
glycolipid transporter activity glycolipid binding phosphatidylinositol-4-phosphate binding ceramide binding
Process(es)
lipid transport protein transport ER to Golgi ceramide transport glycolipid transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0