Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PHOX2B Antibodies

NeoBiotechnologies

PHOX2B Recombinant Rabbit Monoclonal Antibody (PHOX2B/7161R)

View all (13) PHOX2B antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PHOX2B Recombinant Rabbit Monoclonal Antibody (PHOX2B/7161R)

  • Antibody Testing Data (2)
PHOX2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
PHOX2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PHOX2B Antibody (8929-RBM1-P1ABX) in IHC (P)

Formalin-fixed, paraffin-embedded human renal cell carcinoma stained with PHOX2B Recombinant Rabbit Monoclonal Antibody (PHOX2B/7161R). HIER: Tris/EDTA, pH9.0, 45 min. 2 °: HRP-polymer, 30 min. DAB, 5 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PHOX2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PHOX2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))

Product Details

8929-RBM1-P1ABX

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG, kappa

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

PHOX2B/7161R

Immunogen

Recombinant fragment of human PHOX2B protein (exact sequence is proprietary)

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: NBPhox; neuroblastoma paired-type homeobox protein; Neuroblastoma Phox; paired mesoderm homeobox 2b; Paired mesoderm homeobox protein 2B; Paired-like homeobox 2B; PHOX2B homeodomain protein

View more View less

Gene Aliases: NBLST2; NBPhox; PHOX2B; PMX2B

View more View less

UniProt ID: (Human) Q99453

View more View less

Entrez Gene ID: (Human) 8929

View more View less

Function(s)
RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
Process(es)
neuron migration regulation of respiratory gaseous exchange by neurological system process noradrenergic neuron differentiation noradrenergic neuron development brainstem development transcription from RNA polymerase II promoter negative regulation of cell proliferation glial cell differentiation regulation of gene expression positive regulation of G2/M transition of mitotic cell cycle cell differentiation in hindbrain medullary reticular formation development hindbrain tangential cell migration skeletal muscle cell differentiation negative regulation of neuron differentiation positive regulation of neuron differentiation positive regulation of transcription from RNA polymerase II promoter autonomic nervous system development enteric nervous system development sympathetic nervous system development parasympathetic nervous system development inner ear development efferent axon development in a lateral line nerve respiratory system development retrotrapezoid nucleus neuron differentiation sympathetic ganglion development negative regulation of cell cycle arrest dopaminergic neuron differentiation cellular response to BMP stimulus neural crest cell migration involved in autonomic nervous system development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0