Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PDGF-B Antibodies

NeoBiotechnologies

PDGF beta (PDGFB) Monoclonal Antibody (PDGFB, 3072)

View all (14) PDGF-B antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PDGF beta (PDGFB) Monoclonal Antibody (PDGFB, 3072)

  • Antibody Testing Data (3)
PDGF beta (PDGFB) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
PDGF beta (PDGFB) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PDGF beta (PDGFB) Antibody (5155-MSM2-P1ABX) in IHC (P)

Immunohistochemistry (Paraffin) analysis performed in formalin fixed, paraffin-embedded human colon using PDGF beta (PDGFB) Monoclonal Antibody (Product # 5155-MSM2-P1ABX). Inset: PBS instead of primary antibody, secondary antibody negative control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PDGF beta (PDGFB) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PDGF beta (PDGFB) Antibody in Peptide array (ARRAY)
PDGF beta (PDGFB) Antibody in SDS-PAGE (SDS-PAGE)
PDGF beta (PDGFB) Monoclonal Antibody (PDGFB, 3072)

Product Details

5155-MSM2-P1ABX

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1-2 µg/mL
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2c, kappa

Class

Monoclonal

Type

Antibody

Clone

PDGFB, 3072

Immunogen

Recombinant fragment of human PDGFB protein (around aa 27-158)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Immunohistochemistry (PFA fixed): incubate antibody for 30 min RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.

Target Information

This gene encodes a member of the protein family comprised of both platelet-derived growth factors (PDGF) and vascular endothelial growth factors (VEGF). The encoded preproprotein is proteolytically processed to generate platelet-derived growth factor subunit B, which can homodimerize, or alternatively, heterodimerize with the related platelet-derived growth factor subunit A. These proteins bind and activate PDGF receptor tyrosine kinases, which play a role in a wide range of developmental processes. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 17, at sites where this gene and that for collagen type 1, alpha 1 are located, are associated with dermatofibrosarcoma protuberans, a rare skin tumor. Alternative splicing results in multiple transcript variants.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: becaplermin; PDGF subunit B; PDGF, B chain; PDGF-2; platelet-derived growth factor 2; Platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); Platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Proto-oncogene c-Sis

View more View less

Gene Aliases: c-sis; IBGC5; PDGF-2; PDGF2; PDGFB; SIS; SSV

View more View less

UniProt ID: (Human) P01127

View more View less

Entrez Gene ID: (Human) 5155

View more View less

Function(s)
Ras guanyl-nucleotide exchange factor activity platelet-derived growth factor receptor binding protein binding collagen binding growth factor activity superoxide-generating NADPH oxidase activator activity chemoattractant activity identical protein binding protein homodimerization activity phosphatidylinositol-4,5-bisphosphate 3-kinase activity protein heterodimerization activity platelet-derived growth factor binding
Process(es)
MAPK cascade response to hypoxia embryonic placenta development positive regulation of endothelial cell proliferation monocyte chemotaxis platelet degranulation positive regulation of glomerular filtration DNA replication protein phosphorylation transforming growth factor beta receptor signaling pathway heart development positive regulation of cell proliferation response to wounding negative regulation of phosphatidylinositol biosynthetic process negative regulation of platelet activation positive regulation of gene expression negative regulation of gene expression regulation of phosphatidylinositol 3-kinase signaling positive regulation of phosphatidylinositol 3-kinase signaling positive regulation of smooth muscle cell migration cell growth peptidyl-serine phosphorylation peptidyl-tyrosine phosphorylation hemopoiesis extracellular matrix organization positive regulation of cell migration positive regulation of protein autophosphorylation negative regulation of protein binding activation of protein kinase activity activation of protein kinase B activity response to estradiol response to insulin positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway paracrine signaling wound healing response to drug positive regulation of MAP kinase activity positive regulation of MAPK cascade positive regulation of blood vessel endothelial cell migration positive regulation of GTPase activity positive regulation of phosphatidylinositol 3-kinase activity response to estrogen positive regulation of cyclin-dependent protein serine/threonine kinase activity positive regulation of DNA replication positive regulation of mitotic nuclear division negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated phosphatidylinositol phosphorylation platelet-derived growth factor receptor signaling pathway phosphatidylinositol-mediated signaling positive regulation of fibroblast proliferation positive regulation of smooth muscle cell proliferation positive regulation of peptidyl-tyrosine phosphorylation positive chemotaxis positive regulation of chemotaxis positive regulation of cell division cell chemotaxis positive regulation of protein tyrosine kinase activity positive regulation of ERK1 and ERK2 cascade protein kinase C signaling cellular response to growth factor stimulus cellular response to mycophenolic acid positive regulation of glomerular mesangial cell proliferation metanephric glomerular mesangial cell development reactive oxygen species metabolic process positive regulation of calcium ion import positive regulation of hyaluronan biosynthetic process negative regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of vascular smooth muscle cell proliferation positive regulation of vascular associated smooth muscle cell migration negative regulation of vascular smooth muscle cell differentiation positive regulation of vascular smooth muscle cell dedifferentiation positive regulation of reactive oxygen species metabolic process positive regulation of DNA biosynthetic process positive regulation of metanephric mesenchymal cell migration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-585975bbdc-x5cgg:80/100.66.128.25:80.
git-commit: a17be567db00e29ba041ba55be3678c43488a11a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0