Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • OMG Antibodies

Proteintech

OMG Polyclonal Antibody

2 References
View all (11) OMG antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite OMG Polyclonal Antibody

  • Antibody Testing Data (4)
OMG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
OMG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

OMG Antibody (12701-1-AP) in IHC (P)

Immunohistochemistry of paraffin-embedded human brain tissue slide using 12701-1-AP ( OMG Antibody) at dilution of 1:200 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

OMG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
OMG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
OMG Antibody in Western Blot (WB)
OMG Antibody in Western Blot (WB)
OMG Polyclonal Antibody

Product Details

12701-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
View 2 publications 2 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:500
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

OMG Fusion Protein Ag3210 (92-440 aa encoded by BC018050)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.27 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: SLPAHLPRS LWNMSAANNN IKLLDKSDTA YQWNLKYLDV SKNMLEKVVL IKNTLRSLEV LNLSSNKLWT VPTNMPSKLH IVDLSNNSLT QILPGTLINL TNLTHLYLHN NKFTFIPDQS FDQLFQLQEI TLYNNRWSCD HKQNITYLLK WMMETKAHVI GTPCSTQISS LKEHNMYPTP SGFTSSLFTV SGMQTVDTIN SLSVVTQPKV TKIPKQYRTK ETTFGATLSK DTTFTSTDKA FVPYPEDTST ETINSHEAAA ATLTIHLQDG MVTNTSLTSS TKSSPTPMTL SITSGMPNNF SEMPQQSTTL NLWREETTTN VKTPLPSVAN AWKVNASFLL LLNVVVMLAV (92-440 aa encoded by BC018050)

Target Information

Oligodendrocyte myelin glycoprotein (OMG or OMGP) is a cell adhesion molecule contributing to the interactive process required for myelination in the central nervous system. It is also involved in axonogenesis, neuron projection regeneration, and regulation of collateral sprouting of intact axons in response to injury.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: oligodendrocyte myelin glycoprotein; Oligodendrocyte-myelin glycoprotein

View more View less

Gene Aliases: OMG; OMGP

View more View less

UniProt ID: (Human) P23515, (Mouse) Q63912

View more View less

Entrez Gene ID: (Human) 4974, (Rat) 450224, (Mouse) 18377

View more View less

Function(s)
identical protein binding protein homodimerization activity
Process(es)
cell adhesion axonogenesis negative regulation of axonogenesis neuron projection regeneration regulation of collateral sprouting of intact axon in response to injury central nervous system myelination
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0