Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • NEFM Antibodies

MUbio

Neurofilament 160 kD Monoclonal Antibody (RNF406)

View all (34) NEFM antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Neurofilament 160 kD Monoclonal Antibody (RNF406)

Product Details

MUB1305P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:100-1:500
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:100
-

Immunohistochemistry (Frozen) (IHC (F))

1:50-1:100
-

Flow Cytometry (Flow)

1:50-1:100
-
Product Specifications

Species Reactivity

Guinea pig, Hamster, Human, Non-human primate, Rabbit, Xenopus

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

RNF406

Immunogen

Neurofilament 160 kD

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS

Contains

0.09% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

RNF406 reacts exclusively with the phosphorylated isoform of the160 kD neurofilament protein. RNF406 is suitable for immunoblotting and immunohistochemistry on frozen and paraffin-embedded tissues.

The antibody may be stored at 4°C. For prolonged storage prepare appropriate aliquots and store at or below -20°C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4°C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance or the concentration of the product.

Target Information

Neurofilaments (NFs) are a type of intermediate filament (IF) expressed almost exclusively in neuronal cells, and in those cells most prominently in large axons. NFs, in most vertebrates, are composed of three different polypeptide chains with different molecular weights - neurofilament medium protein (NF-M), high (NF-H) and light protein (NF-L), which share sequence and structural similarity in a coiled-coil core domain, but differ in the length and sequence of their N-termini and more dramatically of their C-termini which in the case of NF-M and NF-H form the flexible extensions that link NFs to each other and to other elements in the cytoplasm. NF-M protein tail-mediated interactions of neurofilaments are critical for size and cytoskeletal architecture of axons, and are mediated, in part, by the highly phosphorylated tail domain of this protein. NF-M phosphorylation and O-GlcNAcylation are regulated reciprocally and affect its translocation and filament formation and function. Antibodies to the various neurofilament subunits are very useful cell type markers since the proteins are among the most abundant of the nervous system, are expressed only in neurons and are biochemically very stable.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 160 kDa neurofilament protein; middle molecular weight neurofilament protein NF-M(2); Neurofilament 3; Neurofilament medium polypeptide; neurofilament protein M; Neurofilament triplet M protein; neurofilament, medium polypeptide; neurofilament, medium polypeptide 150kDa; neurofilament, medium polypeptide S homeolog; neurofilament-3 (150 kD medium); NF-M; NMC

View more View less

Gene Aliases: nef-m; NEF3; NEFM; nefm-a; nefm-b; NF-M; NFM; XELAEV_18020229mg

View more View less

UniProt ID: (Human) P07197

View more View less

Entrez Gene ID: (Human) 4741, (Rabbit) 100009343, (Chinese hamster) 100773854, (Xenopus) 397995

View more View less

Function(s)
structural constituent of cytoskeleton protein binding microtubule binding
Process(es)
neurofilament bundle assembly axon development mitophagy in response to mitochondrial depolarization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0