Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DKASGRHYG AVTCEGCKGF FKRSIRKNLV YSCRGSKDCI INKHHRNRCQ YCRLQRCIAF GMKQDSVQCE RKPIEVSREK SSNCAASTEK IYIRKDLRSP LTATPTFVTD SESTRSTGLL DSGMFMNIHP SGVKTESAVL MTSDKAESCQ GDLSTLANVV TSLANLGKTK DLSQNSNEMS MIESLSNDDT SLCEFQEMQT NGDVSRAFDT LAKALNPGES TACQSSVAGM EGSVHLITGD SSINYTEKEG PLLSDSHVAF RLTMPSPMPE YLNVHYIGES ASRLLFLSMH WALSIPSFQA LGQENSISLV KAYWNELFTL GLAQCWQVMN VATILATFVN CLHNSLQQAE G (118-467 aa encoded by BC040141)
This gene encodes a nuclear hormone receptor characterized by a highly conserved DNA binding domain (DBD), a variable hinge region, and a carboxy-terminal ligand binding domain (LBD) that is typical for all members of the steroid/thyroid hormone receptor superfamily. This protein also belongs to a large family of ligand-inducible transcription factors that regulate gene expression by binding to specific DNA sequences within promoters of target genes. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80-3 cNDA; early embryonic nuclear receptor; mTR2; nr2c1; Nuclear receptor subfamily 2 group C member 1; nuclear receptor subfamily 2, group C isoform; nuclear receptor subfamily 2, group C, member 1; nuclear receptor subfamily 2, group H, member 1; Orphan nuclear receptor TR2; orphan receptor, TR2-11; pregnancy specific beta-1-glycoprotein 4; Testicular receptor 2; TR2 nuclear hormone receptor
Gene Aliases: 4831444H07Rik; 80.3; Eenr; NR2C1; Psg4; TR2; TR2-11
UniProt ID: (Human) P13056, (Mouse) Q505F1, (Rat) Q8VIJ4
Entrez Gene ID: (Human) 7181, (Mouse) 22025, (Rat) 252924
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support