Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • NFkB p65 Antibodies

Proteintech

NF-KB p65 Monoclonal Antibody (1B12D11), CoraLite® Plus 488

View all (149) NFkB p65 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite NF-KB p65 Monoclonal Antibody (1B12D11), CoraLite® Plus 488

  • Antibody Testing Data (1)
NF-KB p65 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
NF-KB p65 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

NF-KB p65 Antibody (CL488-66535) in ICC/IF

Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CL488-66535 (NF-kappa B p65 antibody) at dilution of 1:100 {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

NF-KB p65 Antibody in Immunocytochemistry (ICC/IF)

Product Details

CL488-66535

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1:50-1:500
-
Product Specifications

Species Reactivity

Human, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

1B12D11

Immunogen

Recombinant Protein

Conjugate

CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
  • Unconjugated
  • CoraLite 594
  • Request custom conjugation

Excitation/Emission Max

493/522 nm

Form

Liquid

Concentration

2 mg/mL

Purification

Protein G

Storage buffer

PBS, pH 7.3, with 0.5% BSA, 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, store in dark

Shipping conditions

Wet ice

Target Information

Nuclear factor kB p65 (NF-kB p65) is encoded by the RELA gene and is present on chromosome 11 in humans. NF-kB P65 is also known as RelA (v-rel avian reticuloendotheliosis viral oncogene homolog A) and belongs to the Rel family of proteins. It is one of the two subunits of NF-kB (Nuclear factor kappa-light-chain-enhancer of activated B cells) that heterodimerizes with the other subunits p50 or p52. However, binding of TNF alpha to its cognate receptor phosphorylates IKK which in turn phosphorylates I kB allowing proteasomal degradation of I kB. It specifically plays a key role in transcription of immunoglobulin k (kappa) gene in mature B-lymphoid cells.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: EBP-1; MGC131774; NF KB; nf-kappa-b p65; NF-kappa-B p65delta3; NF-kappa-B transcription factor p65; NF-kB p65; NFkappaB p65; nfkb rela; NFkB-p50; NFKB1; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit p65; Nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; REL A; Transcription factor p65; v-rel avian reticuloendotheliosis viral oncogene homolog A; v-rel reticuloendotheliosis viral oncogene homolog A

View more View less

Gene Aliases: NFkB; NFKB3; nos2; p65; RELA

View more View less

UniProt ID: (Human) Q04206, (Rat) Q7TQN4

View more View less

Entrez Gene ID: (Human) 5970, (Rat) 309165

View more View less

Function(s)
RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II distal enhancer sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding protein binding transcription factor binding protein kinase binding chromatin DNA binding ubiquitin protein ligase binding protein complex binding activating transcription factor binding phosphate ion binding identical protein binding protein homodimerization activity actinin binding histone deacetylase binding transcription regulatory region DNA binding protein heterodimerization activity protein N-terminus binding NF-kappaB binding repressing transcription factor binding ankyrin repeat binding RNA polymerase II transcription factor activity, sequence-specific DNA binding enzyme binding sequence-specific DNA binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter liver development hair follicle development stimulatory C-type lectin receptor signaling pathway acetaldehyde metabolic process transcription from RNA polymerase II promoter inflammatory response cellular defense response aging positive regulation of cell proliferation organ morphogenesis response to organic substance response to UV-B positive regulation of Schwann cell differentiation cytokine-mediated signaling pathway membrane protein intracellular domain proteolysis positive regulation of chondrocyte differentiation positive regulation of type I interferon production response to muramyl dipeptide response to progesterone response to insulin response to cobalamin cellular response to hepatocyte growth factor stimulus response to muscle stretch Fc-epsilon receptor signaling pathway negative regulation of protein catabolic process response to drug negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling response to amino acid response to morphine positive regulation of interleukin-12 biosynthetic process negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of insulin receptor signaling pathway regulation of inflammatory response T cell receptor signaling pathway positive regulation of NF-kappaB transcription factor activity response to cAMP defense response to virus cellular response to hydrogen peroxide nucleotide-binding oligomerization domain containing 2 signaling pathway response to interleukin-1 cellular response to lipopolysaccharide cellular response to nicotine cellular response to interleukin-1 cellular response to interleukin-6 cellular response to tumor necrosis factor cellular response to peptide hormone stimulus negative regulation of NIK/NF-kappaB signaling positive regulation of miRNA metabolic process negative regulation of extrinsic apoptotic signaling pathway regulation of transcription from RNA polymerase II promoter defense response I-kappaB kinase/NF-kappaB signaling response to mechanical stimulus response to bacterium response to inorganic substance response to organic cyclic compound response to lipopolysaccharide response to cytokine NIK/NF-kappaB signaling response to hydrogen peroxide innate immune response
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0