Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • NCK1 Antibodies

Abnova

NCK1 Polyclonal Antibody, MaxPab™

View all (13) NCK1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite NCK1 Polyclonal Antibody, MaxPab™

  • Antibody Testing Data (1)
NCK1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
NCK1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

NCK1 Antibody (H00004690-B01P) in WB

NCK1 MaxPab polyclonal antibody. Western Blot analysis of NCK1 expression in A-431. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

NCK1 Antibody in Western Blot (WB)
NCK1 Polyclonal Antibody, MaxPab™

Product Details

H00004690-B01P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

Immunocytochemistry (ICC/IF)

10-100 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG

Class

Polyclonal

Type

Antibody

Immunogen

NCK1 (NP_006144.1, 1 a.a. approximately 377 a.a) full-length human protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Product may be used with Western Blot (Cell lysate).

The Isotype of this product is composed of an IgG Mixture.

Immunogen sequence: MAEEVVVVAK FDYVAQQEQE LDIKKNERLW LLDDSKSWWR VRNSMNKTGF VPSNYVERKN SARKASIVKN LKDTLGIGKV KRKPSVPDSA SPADDSFVDP GERLYDLNMP AYVKFNYMAE REDELSLIKG TKVIVMEKCS DGWWRGSYNG QVGWFPSNYV TEEGDSPLGD HVGSLSEKLA AVVNNLNTGQ VLHVVQALYP FSSSNDEELN FEKGDVMDVI EKPENDPEWW KCRKINGMVG LVPKNYVTVM QNNPLTSGLE PSPPQCDYIR PSLTGKFAGN PWYYGKVTRH QAEMALNERG HEGDFLIRDS ESSPNDFSVS LKAQGKNKHF KVQLKETVYC IGQRKFSTME ELVEHYKKAP IFTSEQGEKL YLVKHLS

Target Information

NCK1 is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Cytoplasmic protein NCK1; melanoma NCK protein; MGC12668; NCK adapter protein 1; NCK tyrosine kinase; Nck-1; non-catalytic region of tyrosine kinase; SH2/SH3 adapter protein NCK-alpha; SH2/SH3 adapter protein NCK1; SH2/SH3 adaptor protein NCK-alpha

View more View less

Gene Aliases: NCK; nck-1; NCK1; NCKalpha

View more View less

UniProt ID: (Human) P16333

View more View less

Entrez Gene ID: (Human) 4690

View more View less

Function(s)
protein kinase inhibitor activity SH3/SH2 adaptor activity ARF guanyl-nucleotide exchange factor activity receptor binding protein binding cytoskeletal adaptor activity protein domain specific binding receptor signaling complex scaffold activity protein binding, bridging receptor tyrosine kinase binding eukaryotic initiation factor eIF2 binding cadherin binding involved in cell-cell adhesion
Process(es)
negative regulation of protein kinase activity substrate-dependent cell migration, cell extension actin filament organization signal complex assembly vesicle-mediated transport lamellipodium assembly positive regulation of actin filament polymerization negative regulation of peptidyl-serine phosphorylation positive regulation of translation in response to endoplasmic reticulum stress Fc-gamma receptor signaling pathway involved in phagocytosis positive regulation of T cell proliferation T cell activation positive regulation of GTPase activity positive regulation of transcription from RNA polymerase II promoter vascular endothelial growth factor receptor signaling pathway T cell receptor signaling pathway response to other organism negative regulation of cell death peptidyl-serine dephosphorylation positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway positive regulation of cap-dependent translational initiation positive regulation of cap-independent translational initiation negative regulation of PERK-mediated unfolded protein response negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0