Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Myogenin Antibodies

Invitrogen

Myogenin Monoclonal Antibody (F5D), Alexa Fluor™ 488, eBioscience™

Advanced Verification
5 Published Figures
4 References
View all (36) Myogenin antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Myogenin Monoclonal Antibody (F5D), Alexa Fluor™ 488, eBioscience™

  • Antibody Testing Data (2)
  • Published Figures (5)
  • Advanced Verification (1)
Myogenin Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Myogenin Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 8

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Myogenin Antibody (53-5643-82) in ICC/IF

Immunofluorescence analysis of MYOG was performed using 70% confluent log phase RD and HeLa cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with Myogenin Monoclonal Antibody (F5D), Alexa Fluor™ 488, eBioscience™ (Product # 53-5643-82, 5 µg/mL) in 0.1% BSA, incubated at 4 degree celsius overnight (Panel a: Green). Nuclei (Panel b: Blue) were stained with ProLong™ Diamond Antifade Mountant ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Myogenin Antibody in Immunocytochemistry (ICC/IF)
Myogenin Antibody in Immunocytochemistry (ICC/IF)
Myogenin Antibody in Immunocytochemistry (ICC/IF)
Myogenin Antibody in Immunocytochemistry (ICC/IF)
Myogenin Antibody in Immunocytochemistry (ICC/IF)
Myogenin Antibody in Western Blot (WB)
Myogenin Antibody in Western Blot (WB)
Myogenin Antibody

Product Details

53-5643-82

Applications
Tested Dilution
Publications

Western Blot (WB)

-
View 2 publications 2 publications

Immunocytochemistry (ICC/IF)

5-20 µg/mL
View 2 publications 2 publications
Product Specifications

Species Reactivity

Dog, Human, Mouse, Rat

Published species

Mouse

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

F5D

Conjugate

Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
  • Unconjugated
  • Request custom conjugation

Excitation/Emission Max

499/520 nm View spectra spectra

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_1907433

Product Specific Information

Description: This F5D monoclonal antibody reacts with human, mouse, dog, and rat myogenin, a 34-kDa transcription factor. Expressed in skeletal and heart muscle, myogenin is a member of the MyoD family of basic-helix-loop-helix proteins, which also includes MyoD, Myf5, and MRF4. This transcription factor interacts with other helix-loop-helix proteins, which may or may not be muscle-specific. Myogenin plays a significant role in myogenic differentiation, even directing nonmuscle cells to the myogenic lineage. Transforming growth factor-beta (TGFb) and bone morphogenetic protein-2 (BMP2) inhibit myogenin transcriptional activity. Predominantly residing within the nucleus, the subcellular localization of myogenin has been shown to be dependent on differentiation status and cell density. For instance, trafficking of myogenin between the nucleus and cytoplasm has been reported during skeletal muscle differentiation to mediate transcription control.

Applications Reported: This F5D antibody has been reported for use in immunocytochemistry.

Applications Tested: This F5D antibody has been tested by immunocytochemistry of the C2C12 cell line. This can be used at less than or equal to 5-20 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.

Filtration: 0.2 µm post-manufacturing filtered.

Target Information

Myogenin is a muscle-specific transcription factor that can induce myogenesis in a variety of cell types in tissue culture. It is a member of a large family of proteins related by sequence homology, the helix-loop-helix proteins. It is essential for the development of functional skeletal muscle.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: bHLHc3; Class C basic helix-loop-helix protein 3; Myf-4; MYOD1-related protein; Myogenic factor 4; Myogenic factor-4; Myogenin

View more View less

Gene Aliases: BHLHC3; myf-4; MYF4; myo; MYOG

View more View less

UniProt ID: (Human) P15173, (Mouse) P12979, (Rat) P20428

View more View less

Entrez Gene ID: (Dog) 490224, (Human) 4656, (Mouse) 17928, (Rat) 29148

View more View less

Function(s)
RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding core promoter binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding transcription factor activity, sequence-specific DNA binding transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding protein binding chromatin DNA binding sequence-specific DNA binding protein heterodimerization activity E-box binding core promoter proximal region sequence-specific DNA binding DNA binding protein dimerization activity molecular_function
Process(es)
ossification transcription, DNA-templated regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter cell cycle multicellular organism development muscle organ development skeletal muscle tissue development negative regulation of cell proliferation positive regulation of myotube differentiation positive regulation of muscle atrophy regulation of skeletal muscle satellite cell proliferation response to muscle activity involved in regulation of muscle adaptation response to electrical stimulus involved in regulation of muscle adaptation striated muscle atrophy response to denervation involved in regulation of muscle adaptation myotube differentiation cell differentiation skeletal muscle cell differentiation mRNA transcription from RNA polymerase II promoter histone H3 acetylation histone H4 acetylation positive regulation of myoblast differentiation negative regulation of glycolytic process positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter skeletal muscle fiber development positive regulation of skeletal muscle fiber development positive regulation of cell cycle arrest cellular response to lithium ion cellular response to growth factor stimulus cellular response to estradiol stimulus regulation of myoblast fusion positive regulation of myoblast fusion positive regulation of oxidative phosphorylation response to gravity skeletal muscle atrophy response to muscle activity skeletal muscle tissue regeneration myoblast differentiation cellular response to magnetism cellular response to retinoic acid muscle cell fate commitment positive regulation of muscle cell differentiation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0