Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • C11orf9 Antibodies

Bioss

MYRF/C11orf9 Polyclonal Antibody

View all (4) C11orf9 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MYRF/C11orf9 Polyclonal Antibody

  • Antibody Testing Data (2)
MYRF/C11orf9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
MYRF/C11orf9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MYRF/C11orf9 Antibody (BS-11191R) in IHC (P)

Formalin-fixed and paraffin embedded rat brain labeled with Anti-MRF/C11orf9 Polyclonal Antibody, Unconjugated (bs-11191R) at 1:200 followed by conjugation to the secondary antibody and DAB staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MYRF/C11orf9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MYRF/C11orf9 Antibody in Western Blot (WB)
MYRF/C11orf9 Polyclonal Antibody

Product Details

BS-11191R

Applications
Tested Dilution
Publications

Western Blot (WB)

1:200
-

Immunohistochemistry (Paraffin) (IHC (P))

1:200
-

Immunohistochemistry (Frozen) (IHC (F))

1:100-1:500
-

Immunocytochemistry (ICC/IF)

1:100-1:500
-

ELISA (ELISA)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic peptide derived from human MYRF, amino acids 401-500.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

Contains

0.02% ProClin 300

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

MYRF is a protein coding gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Myelin gene regulatory factor; Myelin regulatory factor

View more View less

Gene Aliases: 6030439E18; C11orf9; Gm1804; Gm98; KIAA0954; MRF; MYRF; Ndt80; pqn-47; RGD1306622

View more View less

UniProt ID: (Human) Q9Y2G1, (Mouse) Q3UR85

View more View less

Entrez Gene ID: (Human) 745, (Mouse) 225908, (Rat) 293736

View more View less

Function(s)
DNA binding transcription factor activity, sequence-specific DNA binding peptidase activity cysteine-type peptidase activity hydrolase activity sequence-specific DNA binding
Process(es)
transcription, DNA-templated regulation of transcription, DNA-templated proteolysis oligodendrocyte development central nervous system myelination cell differentiation positive regulation of myelination central nervous system myelin maintenance positive regulation of transcription, DNA-templated oligodendrocyte differentiation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0