Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MGST1 Antibodies

Invitrogen

MGST1 Polyclonal Antibody

Advanced Verification
View all (10) MGST1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MGST1 Polyclonal Antibody

  • Antibody Testing Data (4)
  • Advanced Verification (1)
MGST1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
MGST1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MGST1 Antibody (PA5-60845) in ICC/IF

Immunofluorecent analysis of MGST1 in human cell line A-431 using MGST1 Polyclonal Antibody (Product # PA5-60845). Staining shows localization to mitochondria. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MGST1 Antibody in Immunocytochemistry (ICC/IF)
MGST1 Antibody in Immunohistochemistry (IHC)
MGST1 Antibody in Immunohistochemistry (IHC)
MGST1 Antibody in Immunohistochemistry (IHC)
MGST1 Antibody
MGST1 Polyclonal Antibody

Product Details

PA5-60845

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:200-1:500
-

Immunocytochemistry (ICC/IF)

0.25-2 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant protein corresponding to Human MGST1. Recombinant protein control fragment (Product #RP-100835).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.1 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.2, with 40% glycerol

Contains

0.02% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2643943

Product Specific Information

Immunogen sequence: FYRLTRKVFA NPEDCVAFGK GENAKKYLRT DDR

Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 79%.

Target Information

The MAPEG (Membrane Associated Proteins in Eicosanoid and Glutathione metabolism) family consists of six human proteins, two of which are involved in the production of leukotrienes and prostaglandin E, important mediators of inflammation. Other family members, demonstrating glutathione S-transferase and peroxidase activities, are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. This gene encodes a protein that catalyzes the conjugation of glutathione to electrophiles and the reduction of lipid hydroperoxides. This protein is localized to the endoplasmic reticulum and outer mitochondrial membrane where it is thought to protect these membranes from oxidative stress.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: glutathione S-transferase 12; MGC14525; Microsomal glutathione S-transferase 1; Microsomal GST-1; Microsomal GST-I

View more View less

Gene Aliases: GST12; MGST; MGST-I; MGST1

View more View less

UniProt ID: (Human) P10620

View more View less

Entrez Gene ID: (Human) 4257

View more View less

Function(s)
glutathione transferase activity glutathione peroxidase activity protein binding protein homodimerization activity glutathione binding
Process(es)
glutathione metabolic process xenobiotic metabolic process response to organonitrogen compound response to lipopolysaccharide Leydig cell differentiation response to drug oxidation-reduction process protein homotrimerization cellular response to lipid hydroperoxide cellular oxidant detoxification glutathione derivative biosynthetic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0