Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MBLAC2 Antibodies

Invitrogen

MBLAC2 Polyclonal Antibody

View all (3) MBLAC2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MBLAC2 Polyclonal Antibody

  • Antibody Testing Data (1)
MBLAC2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
MBLAC2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MBLAC2 Antibody (PA5-66316) in ICC/IF

Immunofluorescent staining of MBLAC2 in human cell line SK-MEL-30 shows localization to nuclear bodies. Samples were probed using a MBLAC2 Polyclonal Antibody (Product # PA5-66316). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MBLAC2 Antibody in Immunocytochemistry (ICC/IF)
MBLAC2 Polyclonal Antibody

Product Details

PA5-66316

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

0.25-2 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant Human MBLAC2
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.1 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.2, with 40% glycerol

Contains

0.02% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2663641

Product Specific Information

Immunogen sequence: MSALEWYAHKS LGDGIFWIQE RFYESGNRAN IWLVRGSEQD VVIDTGLGLR SLPEYLYSSG LLQDREAKED A

Highest antigen sequence identity to the following orthologs - mouse 96%, rat 97%.

Target Information

MBLAC2 is a protein coding gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Acyl-CoA thioesterase MBLAC2; Acyl-coenzyme A thioesterase MBLAC2; Beta-lactamase MBLAC2; Metallo-beta-lactamase domain-containing protein 2; Palmitoyl-coenzyme A thioesterase MBLAC2

View more View less

Gene Aliases: MBLAC2

View more View less

UniProt ID: (Human) Q68D91

View more View less

Entrez Gene ID: (Human) 153364

View more View less

Function(s)
molecular_function hydrolase activity metal ion binding
Process(es)
biological_process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0