Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MAGOHB Antibodies

Invitrogen

MAGOHB Polyclonal Antibody

View all (4) MAGOHB antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MAGOHB Polyclonal Antibody

  • Antibody Testing Data (1)
MAGOHB Antibody in Western Blot (WB)
Group 53 Created with Sketch.
MAGOHB Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MAGOHB Antibody (PA5-116666) in WB

Western blot analysis of MAGOHB in extracts of various cell lines. Samples were incubated with MAGOHB Polyclonal antibody (Product # PA5-116666) using a dilution of 1:3,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 90s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MAGOHB Antibody in Western Blot (WB)
MAGOHB Polyclonal Antibody

Product Details

PA5-116666

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human MAGOHB (NP_0605181)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.56 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.01% thimerosal

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

RRID

AB_2901297

Product Specific Information

Positive Samples: HeLa, Jurkat, PC-12, U-87MG

Immunogen sequence: MAVASDFYLR YYVGHKGKFG HEFLEFEFRP DGKLRYANNS NYKNDVMIRK EAYVHKSVME ELKRIIDDSE ITKEDDALWP PPDRVGRQEL EIVIGDEHIS FTTSKIGSLI DVNQSKDPEG LRVFYYLVQD LKCLVFSLIG LHFKIKPI

Target Information

MAGOHB is a protein coding gene involved in mRNA splicing and in the nonsense-mediated decay (NMD) pathway.

⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: mago-nashi homolog B; Protein mago nashi homolog 2

View more View less

Gene Aliases: mago; magoh; MAGOH2; MAGOHB; MGN2

View more View less

UniProt ID: (Human) Q96A72

View more View less

Entrez Gene ID: (Human) 55110, (Rat) 690303

View more View less

Function(s)
protein binding poly(A) RNA binding
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay mRNA processing RNA splicing mRNA transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0