Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MAGI1 Antibodies

Abnova

MAGI1 Polyclonal Antibody, MaxPab™

View all (10) MAGI1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MAGI1 Polyclonal Antibody, MaxPab™

  • Antibody Testing Data (1)
MAGI1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
MAGI1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MAGI1 Antibody (H00009223-B01P) in WB

Western Blot analysis of MAGI1 expression in transfected 293T cell line by MAGI1 MaxPab polyclonal antibody. Lane 1: MAGI1 transfected lysate(138.2 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MAGI1 Antibody in Western Blot (WB)
MAGI1 Polyclonal Antibody, MaxPab™

Product Details

H00009223-B01P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG

Class

Polyclonal

Type

Antibody

Immunogen

MAGI1 (AAI67863.1, 1 a.a. approximately 1256 a.a) full-length human protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5-1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Product may be used with Western Blot (Transfected lysate).

The Isotype of this product is composed of an IgG Mixture.

Immunogen sequence: MSKVIQKKNH WTSRVHECTV KRGPQGELGV TVLGGAEHGE FPYVGAVAAV EAAGLPGGGE GPRLGEGELL LEVQGVRVSG LPRYDVLGVI DSCKEAVTFK AVRQGGRLNK DLRHFLNQRF QKGSPDHELQ QTIRDNLYRH AVPCTTRSPR EGEVPGVDYN FLTVKEFLDL EQSGTLLEVG TYEGNYYGTP KPPSQPVSGK VITTDALHSL QSGSKQSTPK RTKSYNDMQN AGIVHAENEE EDDVPEMNSS FTADSGEQEE HTLQETALPP VNSSIIAAPI TDPSQKFPQY LPLSAEDNLG PLPENWEMAY TENGEVYFID HNTKTTSWLD PRCLNKQQKP LEECEDDEGV HTEELDSELE LPAGWEKIED PVYGIYYVDH INRKTQYENP VLEAKRKKQL EQQQQQQQQQ QQQQQQQQQQ QTEEWTEDHS ALVPPVIPNH PPSNPEPARE VPLQGKPFFT RNPSELKGKF IHTKLRKSSR GFGFTVVGGD EPDEFLQIKS LVLDGPAALD GKMETGDVIV SVNDTCVLGH THAQVVKIFQ SIPIGASVDL ELCRGYPLPF DPDDPNTSLV TSVAILDKEP IIVNGQETYD SPASHSSKTG KVNGMKDARP SSPADVASNS SHGYPNDTVS LASSIATQPE LITVHIVKGP MGFGFTIADS PGGGGQRVKQ IVDSPRCRGL KEGDLIVEVN KKNVQALTHN QVVDMLVECP KGSEVTLLVQ RGGLPVPKKS PKSQPLERKD SQNSSQHSVS SHRSLHTASP SHSTQVLPEF PPAEAQAPDQ TDSSGQKKPD PFKIWAQSRS MYENRPMSPS PASGLSKGER EREINSTNFG ECPIPDYQEQ DIFLWRKETG FGFRILGGNE PGEPIYIGHI VPLGAADTDG RLRSGDELIC VDGTPVIGKS HQLVVQLMQQ AAKQGHVNLT VRRKVVFAVP KTENEVPSPA SSHHSSNQPA SLTEEKRTPQ GSQNSLNTVS SGSGSTSGIG SGGGGGSGVV STVVQPYDVE IRRGENEGFG FVIVSSVSRP EAGTTFAGNA CVAMPHKIGR IIEGSPADRC GKLKVGDRIL AVNGCSITNK SHSDIVNLIK EAGNTVTLRI IPGDESSNAT LLTNAEKIAT ITTTHTPSQQ GTQETRNTTK PKQESQFEFK APQATQEQDF YTVELERGAK GFGFSLRGGR EYNMDLYVLR LAEDGPAERC GKMRIGDEIL EINGETTKNM KHSRAIELIK NGGRRVRLFL KRGDGSVPEY GGSNYENIPS FPGMTP

Target Information

The protein encoded by this gene is a member of the membrane-associated guanylate kinase homologue (MAGUK) family. MAGUK proteins participate in the assembly of multiprotein complexes on the inner surface of the plasma membrane at regions of cell-cell contact. The product of this gene may play a role as scaffolding protein at cell-cell junctions. Alternatively spliced transcript variants encoding different isoforms have been identified.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: AIP-3; Atrophin-1-interacting protein 3; BAI1-associated protein 1; BAP-1; MAGI-1; Membrane-associated guanylate kinase inverted 1; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 1; Trinucleotide repeat-containing gene 19 protein; WW domain-containing protein 3; WWP3

View more View less

Gene Aliases: AIP-3; AIP3; BAIAP1; BAP-1; BAP1; MAGI-1; MAGI1; Magi1d; TNRC19; WWP3

View more View less

UniProt ID: (Human) Q96QZ7

View more View less

Entrez Gene ID: (Human) 9223

View more View less

Function(s)
protein binding ATP binding protein C-terminus binding protein complex scaffold alpha-actinin binding
Process(es)
protein complex assembly cell adhesion cell surface receptor signaling pathway neuron death
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0