Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • LPAM-1 Antibodies

Proteintech

LPAM-1 Monoclonal Antibody (DATK32), APC

View all (13) LPAM-1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite LPAM-1 Monoclonal Antibody (DATK32), APC

  • Antibody Testing Data (2)
LPAM-1 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
LPAM-1 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

LPAM-1 Antibody (APC-65101-100UG) in Flow

1X10^6 C57BL/6 Mouse bone marrow cells were surface stained with 0.25 µg FITC-Anti-Human/Mouse CD45R (B220) (FITC-65139, clone RA3-6B2) and 0.25 µg APC-Anti-Mouse LPAM-1 (Product # APC-65101-100UG), clone DATK32). Cells were not fixed. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

LPAM-1 Antibody in Flow Cytometry (Flow)
LPAM-1 Antibody in Flow Cytometry (Flow)
LPAM-1 Monoclonal Antibody (DATK32), APC

Product Details

APC-65101-100UG

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

Assay-dependent
-
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Rat / IgG2a

Class

Monoclonal

Type

Antibody

Clone

DATK32

Conjugate

APC APC APC
  • Unconjugated
  • CoraLite 594
  • PE 
  • Request custom conjugation

Excitation/Emission Max

651/660 nm View spectra spectra

Form

Liquid

Concentration

0.2 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2, with 0.1% gelatin

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark

Shipping conditions

Wet ice

Target Information

Integrin beta 7 associates with integrin alpha 4 (CD49d) to form the alpha 4 beta 7 integrin LPAM-1, expressed on intraepithelial lymphocytes. It also associates with alpha E (CD103) to form the alpha E beta 7 integrin HML-1, expressed on T cells adjacent to mucosal epithelium and intraepithelial lymphocytes. Ligands for LPAM-1 include VCAM-1 (CD106), MAdCAM-1 and fibronectin, while the ligand of HML-1 is E-cadherin (CD324). Integrin beta 7 plays an important role in the adhesion of leukocytes to endothelial cells promoting the transmigration of leukocytes to extravascular spaces during the inflammatory response.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD49 antigen-like family member D; CD49d; Integrin alpha-4; Integrin alpha-IV; integrin alpha4B; Integrin beta-7; Integrin beta-P; LPAM subunit alpha; Lymphocyte Peyer patch adhesion molecules subunit alpha; M290 IEL antigen; VLA; VLA 4; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha; VLA4 subunit alpha

View more View less

Gene Aliases: CD49D; Itga4; Itga4B; Itgb7; Ly69

View more View less

UniProt ID: (Mouse) Q00651, (Mouse) P26011

View more View less

Entrez Gene ID: (Mouse) 16401, (Mouse) 16421

View more View less

Function(s)
fibronectin binding metal ion binding cell adhesion molecule binding protein antigen binding receptor activity
Process(es)
cell-matrix adhesion involved in ameboidal cell migration cell adhesion cell-matrix adhesion integrin-mediated signaling pathway heterotypic cell-cell adhesion substrate adhesion-dependent cell spreading receptor clustering leukocyte migration leukocyte tethering or rolling T cell migration blood vessel remodeling positive regulation of leukocyte migration heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules leukocyte cell-cell adhesion heart development single organismal cell-cell adhesion cell migration diapedesis face development mesenchyme development chorio-allantoic fusion visceral serous pericardium development negative regulation of protein homodimerization activity import into cell positive regulation of leukocyte cell-cell adhesion positive regulation of leukocyte tethering or rolling positive regulation of T cell migration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-585975bbdc-x5cgg:80/100.66.128.25:80.
git-commit: a17be567db00e29ba041ba55be3678c43488a11a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0