Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • LIMD1 Antibodies

Invitrogen

LIMD1 Polyclonal Antibody

View all (10) LIMD1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite LIMD1 Polyclonal Antibody

  • Antibody Testing Data (4)
LIMD1 Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.
LIMD1 Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

LIMD1 Antibody (PA5-43594) in IHC

Immunohistochemistry analysis of human placenta tissue using an anti-LIMD1 polyclonal antibody (Product # PA5-43594). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

LIMD1 Antibody in Immunohistochemistry (IHC)
LIMD1 Antibody in Immunohistochemistry (IHC)
LIMD1 Antibody in Immunohistochemistry (IHC)
LIMD1 Antibody in Western Blot (WB)
LIMD1 Polyclonal Antibody

Product Details

PA5-43594

Applications
Tested Dilution
Publications

Western Blot (WB)

1 µg/mL
-

Immunohistochemistry (IHC)

5 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthetic peptide directed towards the N-terminal of human LIMD1 (aa 1-50).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS with 2% sucrose

Contains

0.09% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2576727

Product Specific Information

Peptide sequence: MDKYDDLGLE ASKFIEDLNM YEASKDGLFR VDKGAGNNPE FEETRRVFAT

Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 100%

Target Information

The Zyxin family of proteins contains five members: Ajuba, Limd1, LPP, TRIP6 and Zyxin. Limd1 (LIM domain-containing protein 1) is a ubiquitously expressed tumor suppressor containing three LIM zinc-binding domains. LIM domains consist of a cysteine-rich consensus sequence containing two distinct zinc-binding subdomains, which mediate protein-protein interactions. Limd1 interacts with the proteins SQSTM1, Rb, p62 and TRAF6. Limd1 was first identified when the deletion of its gene was noted in some cervical cancers. Limd1 blocks in vitro and in vivo tumor growth and is downregulated in lung cancer.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: LIM domain-containing protein 1

View more View less

Gene Aliases: LIMD1

View more View less

UniProt ID: (Human) Q9UGP4

View more View less

Entrez Gene ID: (Human) 8994

View more View less

Function(s)
transcription corepressor activity protein binding zinc ion binding
Process(es)
response to hypoxia osteoblast development transcription, DNA-templated regulation of transcription, DNA-templated cytoskeleton organization signal transduction multicellular organism development regulation of cell shape phosphorylation cell migration cytoplasmic mRNA processing body assembly gene silencing by miRNA negative regulation of hippo signaling negative regulation of osteoblast differentiation negative regulation of transcription, DNA-templated negative regulation of canonical Wnt signaling pathway positive regulation of gene silencing by miRNA
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0