Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • LIMK2 Antibodies

Fabgennix

LIM kinase 2 Polyclonal Antibody, FITC

View all (31) LIMK2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite LIM kinase 2 Polyclonal Antibody, FITC

Product Details

LIMK2-Y-FITC

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

Immunohistochemistry (IHC)

1:50-1:250
-

ELISA (ELISA)

1:10,000
-

Immunoprecipitation (IP)

1:50-1:250
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Chicken / IgY

Class

Polyclonal

Type

Antibody

Immunogen

Synthetic peptide taken within amino acid region 570-640 on LIMK2 protein.

Conjugate

FITC FITC FITC

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

0.5-1.5 mg/mL

Purification

Affinity chromatography

Storage buffer

proprietary buffer, pH 7.4-7.8, with 30% glycerol, 0.5% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C, store in dark

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

LIMK2 is a serine/threonine kinase involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin. LIMK2 has also been implicated in brain development. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. The protein encoded by LIMK2 is phosphorylated and activated by ROCK, a downstream effector of Rho, and the encoded protein, in turn, phosphorylates cofilin, inhibiting its actin-depolymerizing activity. It is thought that this pathway contributes to Rho-induced reorganization of the actin cytoskeleton. At least three transcript variants encoding different isoforms have been found for LIMK2.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: EC 2.7.11.1; kinase LIMK2; LIM domain kinase 2; LIM kinase 2; LIM motif-containing protein kinase 2; LIMK; LIMK 2; LIMK-1; LIMK-2; LIMK1; LINhp-2; OTTHUMP00000199483

View more View less

Gene Aliases: Limk-2; LIMK2; Limk2b; Link2

View more View less

UniProt ID: (Human) P53671, (Mouse) O54785, (Rat) P53670

View more View less

Entrez Gene ID: (Human) 3985, (Mouse) 16886, (Rat) 29524

View more View less

Function(s)
protein serine/threonine kinase activity protein binding ATP binding zinc ion binding protein heterodimerization activity nucleotide binding protein kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups metal ion binding
Process(es)
protein phosphorylation spermatogenesis phosphorylation spermatid development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0