Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Galectin 3 Antibodies

OriGene

LGALS3 Monoclonal Antibody (OTI1C7), TrueMAB™

View all (131) Galectin 3 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite LGALS3 Monoclonal Antibody (OTI1C7), TrueMAB™

  • Antibody Testing Data (23)
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 23

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

LGALS3 Antibody (CF801621) in IHC (P)

Immunohistochemical staining of paraffin-embedded human lymphoma tissue using anti-LGALS3 mouse monoclonal antibody. (Heat-induced epitope retrieval by 1 mM EDTA in 10mM Tris, pH8.5, 120°C for 3min, TA801621) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGALS3 Antibody in Western Blot (WB)
LGALS3 Monoclonal Antibody (OTI1C7), TrueMAB™

Product Details

CF801621

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG

Class

Monoclonal

Type

Antibody

Clone

OTI1C7

Immunogen

Full length human recombinant protein of human LGALS3 produced in E.coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

Galactose-specific lectin which binds IgE. May mediate with the alpha-3, beta-1 integrin the stimulation by CSPG4 of endothelial cells migration. Together with DMBT1, required for terminal differentiation of columnar epithelial cells during early embryogenesis. In the nucleus: acts as a pre-mRNA splicing factor. Involved in acute inflammatory responses including neutrophil activation and adhesion, chemoattraction of monocytes macrophages, opsonization of apoptotic neutrophils, and activation of mast cells.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 35 kDa lectin; advanced glycation end-product receptor 3; Carbohydrate-binding protein 35; CBP 35; CBP-30; Gal-3; Galactose-specific lectin 3; Galactoside-binding protein; GALBP; Galectin-3; Galectin3; IgE-binding protein; L-29; L-31; Laminin-binding protein; Lectin L-29; lectin, galactoside-binding, soluble, 3; MAC-2 antigen

View more View less

Gene Aliases: CBP35; GAL3; GALBP; GALIG; L31; LGALS2; LGALS3; MAC2

View more View less

UniProt ID: (Human) P17931

View more View less

Entrez Gene ID: (Human) 3958

View more View less

Function(s)
protein binding IgE binding carbohydrate binding chemoattractant activity laminin binding poly(A) RNA binding
Process(es)
monocyte chemotaxis mRNA processing RNA splicing neutrophil chemotaxis epithelial cell differentiation regulation of T cell proliferation innate immune response negative regulation of endocytosis eosinophil chemotaxis macrophage chemotaxis negative regulation of T cell receptor signaling pathway positive chemotaxis regulation of T cell apoptotic process mononuclear cell migration positive regulation of mononuclear cell migration positive regulation of calcium ion import regulation of extrinsic apoptotic signaling pathway via death domain receptors negative regulation of immunological synapse formation negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell positive regulation of dendritic cell differentiation negative regulation of extrinsic apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0