Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Insulin Antibodies

NeoBiotechnologies

Insulin/IRDN (beta-Cell and Insulinoma Marker) Monoclonal Antibody (E2-E3, 2D11-H5 (INS04, INS05))

View all (73) Insulin antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Insulin/IRDN (beta-Cell and Insulinoma Marker) Monoclonal Antibody (E2-E3, 2D11-H5 (INS04, INS05))

Insulin/IRDN (beta-Cell and Insulinoma Marker) Monoclonal Antibody (E2-E3, 2D11-H5 (INS04, INS05))
  • Antibody Testing Data (2)
Insulin/IRDN (beta-Cell and Insulinoma Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Insulin/IRDN (beta-Cell and Insulinoma Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Insulin/IRDN (beta-Cell and Insulinoma Marker) Antibody (3630-MSM3-P0) in IHC (P)

Formalin-fixed, paraffin-embedded human Pancreas stained with Insulin Mouse Monoclonal Antibody (E2-E3+2D11-H5). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Insulin/IRDN (beta-Cell and Insulinoma Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Insulin/IRDN (beta-Cell and Insulinoma Marker) Antibody in SDS-PAGE (SDS-PAGE)

Product Details

3630-MSM3-P0

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-
Product Specifications

Species Reactivity

Bovine, Human, Mouse, Pig, Rabbit

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

E2-E3, 2D11-H5 (INS04, INS05)

Immunogen

Purified pig insulin, conjugated to KLH (E2-E3 & 2D11-H5)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Wet ice

Target Information

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Ins; Insulin; Insulin-1; Insulin-2; preproinsulin; proinsulin

View more View less

Gene Aliases: AA986540; BOS_25069; IDDM; IDDM1; IDDM2; ILPR; INS; Ins-1; Ins-2; Ins1; Ins2; Ins2-rs1; InsII; IRDN; Mody; MODY10; Mody4; proinsulin

View more View less

UniProt ID: (Pig) P01315, (Human) P01308, (Mouse) P01325, (Mouse) P01326

View more View less

Entrez Gene ID: (Pig) 397415, (Bovine) 280829, (Rabbit) 100009181, (Human) 3630, (Mouse) 16333, (Mouse) 16334

View more View less

Function(s)
hormone activity protease binding insulin receptor binding insulin-like growth factor receptor binding protein binding identical protein binding chaperone binding protein complex binding
Process(es)
glucose metabolic process MAPK cascade negative regulation of acute inflammatory response regulation of transcription, DNA-templated regulation of cellular amino acid metabolic process ER to Golgi vesicle-mediated transport acute-phase response G-protein coupled receptor signaling pathway cell-cell signaling positive regulation of cell proliferation insulin receptor signaling pathway positive regulation of gene expression positive regulation of phosphatidylinositol 3-kinase signaling glucose transport regulation of transmembrane transporter activity positive regulation of cell growth positive regulation of cell migration positive regulation of protein autophosphorylation activation of protein kinase B activity positive regulation of cellular protein metabolic process negative regulation of protein oligomerization regulation of protein localization negative regulation of NAD(P)H oxidase activity wound healing negative regulation of protein catabolic process glucose homeostasis positive regulation of MAPK cascade cellular protein metabolic process positive regulation of nitric oxide biosynthetic process positive regulation of cell differentiation negative regulation of gluconeogenesis positive regulation of glycogen biosynthetic process positive regulation of DNA replication negative regulation of glycogen catabolic process positive regulation of glycolytic process positive regulation of mitotic nuclear division negative regulation of proteolysis negative regulation of vasodilation positive regulation of vasodilation negative regulation of fatty acid metabolic process positive regulation of glucose import positive regulation of insulin receptor signaling pathway alpha-beta T cell activation positive regulation of lipid biosynthetic process regulation of protein secretion negative regulation of protein secretion positive regulation of cytokine secretion positive regulation of peptidyl-tyrosine phosphorylation negative regulation of lipid catabolic process positive regulation of nitric-oxide synthase activity positive regulation of NF-kappaB transcription factor activity positive regulation of protein kinase B signaling fatty acid homeostasis negative regulation of respiratory burst involved in inflammatory response positive regulation of respiratory burst positive regulation of peptide hormone secretion positive regulation of brown fat cell differentiation positive regulation of protein localization to nucleus negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway negative regulation of feeding behavior negative regulation of transcription from RNA polymerase II promoter activation of MAPK activity carbohydrate metabolic process ER overload response activation of JUN kinase activity myoblast fusion regulation of gene expression myotube differentiation lipid catabolic process regulation of phosphorylation regulation of protein binding positive regulation of phosphatidylinositol 3-kinase activity canonical Wnt signaling pathway intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c69d76d75-pfrg2:80/100.66.135.80:80.
git-commit: 261285cf70483950061b29b455a88d65ecd945f9
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0