Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • INTS3 Antibodies

Proteintech

INTS3 Polyclonal Antibody

12 References
View all (8) INTS3 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite INTS3 Polyclonal Antibody

  • Antibody Testing Data (10)
INTS3 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
INTS3 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 10

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

INTS3 Antibody (16620-1-AP) in ICC/IF

Immunofluorescent analysis of ( 10% Formaldehyde ) fixed HeLa cells using 16620-1-AP (INTS3 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

INTS3 Antibody in Immunocytochemistry (ICC/IF)
INTS3 Antibody in Immunocytochemistry (ICC/IF)
INTS3 Antibody in Western Blot (WB)
INTS3 Antibody in Western Blot (WB)
INTS3 Antibody in Western Blot (WB)
INTS3 Antibody in Western Blot (WB)
INTS3 Antibody in Western Blot (WB)
INTS3 Antibody in Western Blot (WB)
INTS3 Antibody in Immunoprecipitation (IP)
INTS3 Antibody in Immunoprecipitation (IP)
INTS3 Polyclonal Antibody

Product Details

16620-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:3,000
View 9 publications 9 publications

Immunocytochemistry (ICC/IF)

1:50-1:500
-

Immunoprecipitation (IP)

0.5-4.0 µg
-

ChIP assay (ChIP)

Assay-dependent
View 2 publications 2 publications

Not applicable (N/A)

-
View 1 publication 1 publication
Product Specifications

Species Reactivity

Human, Mouse, Rat

Published species

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

INTS3 Fusion Protein Ag9917 (91-445 aa encoded by BC025254)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.27 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: YHLLYYLRA SKAAAGKMNL YESFAQATQL GDLHTCLMMD MKACQEDDVR LLCHLTPSIY TEFPDETLRS GELLNMIVAV IDSAQLQELV CHVMMGNLVM FRKDSVLNIL IQSLDWETFE QYCAWQLFLA HNIPLETIIP ILQHLKYKEH PEALSCLLLQ LRREKPSEEM VKMVLSRPCH PDDQFTTSIL RHWCMKHDEL LAEHIKSLLI KNNSLPRKRQ SLRSSSSKLA QLTLEQILEH LDNLRLNLTN TKQNFFSQTP ILQALQHVQA SCDEAHKMKF SDLFSLAEEY EDSSTKPPKS RRKAALSSPR SRKNATQPPN AEEESGSSSA SEEEDTKPKP TKRKRKGSSA VGSDSD (91-445 aa encoded by BC025254)

Target Information

Component of the Integrator complex. The Integrator complex is involved in the small nuclear RNAs (snRNA) U1 and U2 transcription and in their 3'-box-dependent processing. The Integrator complex is associated with the C-terminal domain (CTD) of RNA polymerase II largest subunit (POLR2A) and is recruited to the U1 and U2 snRNAs genes.; FUNCTION: Component of the SOSS complex, a multiprotein complex that functions downstream of the MRN complex to promote DNA repair and G2/M checkpoint. The SOSS complex associates with single-stranded DNA at DNA lesions and influences diverse endpoints in the cellular DNA damage response including cell-cycle checkpoint activation, recombinational repair and maintenance of genomic stability. The SOSS complex is required for efficient homologous recombination-dependent repair of double-strand breaks (DSBs) and ATM-dependent signaling pathways. In the SOSS complex, it is required for the assembly of the complex and for stabilization of the complex at DNA damage sites.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: Int3; Integrator complex subunit 3; Sensor of single-strand DNA complex subunit A; sensor of ssDNA subunit A; SOSS complex subunit A; SOSS-A

View more View less

Gene Aliases: C1orf193; C1orf60; C77668; INT3; INTS3; RGD1304593; SOSS-A; SOSSA

View more View less

UniProt ID: (Human) Q68E01, (Mouse) Q7TPD0

View more View less

Entrez Gene ID: (Human) 65123, (Mouse) 229543, (Rat) 361988

View more View less

Function(s)
protein binding molecular_function
Process(es)
DNA repair cellular response to DNA damage stimulus mitotic cell cycle checkpoint response to ionizing radiation snRNA processing snRNA transcription from RNA polymerase II promoter
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0