Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Insulin Antibodies

Proteintech

INS Monoclonal Antibody (4B6A7), CoraLite® 594

View all (73) Insulin antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite INS Monoclonal Antibody (4B6A7), CoraLite® 594

  • Antibody Testing Data (1)
INS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
INS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

INS Antibody (CL59466198100UL) in IHC (P)

Immunofluorescent analysis of (4% PFA) fixed Mouse pancreas tissue using the CoraLite 594-conjugated version of this antibody, (Product # CL59466198100UL) (INS antibody), at dilution of 1:400. The α cells of pancreas was labelled in green with CL488-67286 (Glucagon antibody). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

INS Antibody in Immunohistochemistry (Paraffin) (IHC (P))
INS Monoclonal Antibody (4B6A7), CoraLite® 594

Product Details

CL59466198100UL

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:400
-

Immunocytochemistry (ICC/IF)

1:200-1:800
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

4B6A7

Conjugate

CoraLite® 594 CoraLite® 594 CoraLite® 594
  • Unconjugated
  • CoraLite Plus 488
  • Request custom conjugation

Excitation/Emission Max

593/614 nm

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 0.5% BSA, 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles, store in dark

Shipping conditions

Wet ice

Target Information

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Ins; Insulin; Insulin-1; Insulin-2; preproinsulin; proinsulin

View more View less

Gene Aliases: AA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins-1; Ins-2; Ins1; Ins2; Ins2-rs1; InsII; IRDN; Mody; MODY10; Mody4; proinsulin

View more View less

UniProt ID: (Human) P01308, (Rat) P01322, (Mouse) P01325, (Rat) P01323, (Mouse) P01326

View more View less

Entrez Gene ID: (Human) 3630, (Rat) 24505, (Mouse) 16333, (Rat) 24506, (Mouse) 16334

View more View less

Function(s)
protease binding insulin receptor binding insulin-like growth factor receptor binding hormone activity protein binding identical protein binding chaperone binding protein complex binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter activation of MAPK activity glucose metabolic process ER overload response myoblast fusion biological_process insulin receptor signaling pathway lipid catabolic process regulation of protein binding positive regulation of phosphatidylinositol 3-kinase activity positive regulation of peptidyl-tyrosine phosphorylation canonical Wnt signaling pathway intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress MAPK cascade negative regulation of acute inflammatory response regulation of cellular amino acid metabolic process acute-phase response G-protein coupled receptor signaling pathway positive regulation of cell proliferation response to organic substance positive regulation of phosphatidylinositol 3-kinase signaling glucose transport regulation of transmembrane transporter activity activation of protein kinase B activity positive regulation of cellular protein metabolic process negative regulation of protein oligomerization regulation of protein localization negative regulation of NAD(P)H oxidase activity response to cytokine wound healing negative regulation of protein catabolic process glucose homeostasis positive regulation of MAPK cascade response to peptide hormone positive regulation of glycogen biosynthetic process positive regulation of DNA replication negative regulation of glycogen catabolic process positive regulation of glycolytic process positive regulation of mitotic nuclear division negative regulation of proteolysis negative regulation of fatty acid metabolic process positive regulation of glucose import positive regulation of insulin receptor signaling pathway alpha-beta T cell activation regulation of protein secretion negative regulation of protein secretion positive regulation of cytokine secretion negative regulation of lipid catabolic process positive regulation of NF-kappaB transcription factor activity response to cAMP positive regulation of protein kinase B signaling fatty acid homeostasis negative regulation of respiratory burst involved in inflammatory response positive regulation of respiratory burst positive regulation of protein localization to nucleus negative regulation of feeding behavior regulation of transcription, DNA-templated ER to Golgi vesicle-mediated transport cell-cell signaling positive regulation of gene expression positive regulation of cell growth positive regulation of cell migration positive regulation of protein autophosphorylation cellular protein metabolic process positive regulation of nitric oxide biosynthetic process positive regulation of cell differentiation negative regulation of gluconeogenesis negative regulation of vasodilation positive regulation of vasodilation positive regulation of lipid biosynthetic process positive regulation of nitric-oxide synthase activity positive regulation of peptide hormone secretion positive regulation of brown fat cell differentiation negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway carbohydrate metabolic process activation of JUN kinase activity regulation of gene expression myotube differentiation regulation of phosphorylation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0