Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • IL18RAP Antibodies

Abnova

IL18RAP Polyclonal Antibody, MaxPab™

View all (21) IL18RAP antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite IL18RAP Polyclonal Antibody, MaxPab™

  • Antibody Testing Data (1)
IL18RAP Antibody in Western Blot (WB)
Group 53 Created with Sketch.
IL18RAP Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

IL18RAP Antibody (H00008807-B01P) in WB

Western Blot analysis of IL18RAP expression in transfected 293T cell line by IL18RAP MaxPab polyclonal antibody. Lane 1: IL18RAP transfected lysate(68.30 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

IL18RAP Antibody in Western Blot (WB)
IL18RAP Polyclonal Antibody, MaxPab™

Product Details

H00008807-B01P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG

Class

Polyclonal

Type

Antibody

Immunogen

IL18RAP (NP_003844.1, 1 a.a. approximately 599 a.a) full-length human protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Product may be used with Western Blot (Transfected lysate).

The Isotype of this product is composed of an IgG Mixture.

Immunogen sequence: MLCLGWIFLW LVAGERIKGF NISGCSTKKL LWTYSTRSEE EFVLFCDLPE PQKSHFCHRN RLSPKQVPEH LPFMGSNDLS DVQWYQQPSN GDPLEDIRKS YPHIIQDKCT LHFLTPGVNN SGSYICRPKM IKSPYDVACC VKMILEVKPQ TNASCEYSAS HKQDLLLGST GSISCPSLSC QSDAQSPAVT WYKNGKLLSV ERSNRIVVDE VYDYHQGTYV CDYTQSDTVS SWTVRAVVQV RTIVGDTKLK PDILDPVEDT LEVELGKPLT ISCKARFGFE RVFNPVIKWY IKDSDLEWEV SVPEAKSIKS TLKDEIIERN IILEKVTQRD LRRKFVCFVQ NSIGNTTQSV QLKEKRGVVL LYILLGTIGT LVAVLAASAL LYRHWIEIVL LYRTYQSKDQ TLGDKKDFDA FVSYAKWSSF PSEATSSLSE EHLALSLFPD VLENKYGYSL CLLERDVAPG GVYAEDIVSI IKRSRRGIFI LSPNYVNGPS IFELQAAVNL ALDDQTLKLI LIKFCYFQEP ESLPHLVKKA LRVLPTVTWR GLKSVPPNSR FWAKMRYHMP VKNSQGFTWN QLRITSRIFQ WKGLSRTETT GRSSQPKEW

Target Information

The protein encoded by this gene is an accessory subunit of the heterodimeric receptor for IL18. This protein enhances the IL18 binding activity of IL18R1 (IL1RRP), a ligand binding subunit of IL18 receptor. The coexpression of IL18R1 and this protein is required for the activation of NF-kappaB and MAPK8 (JNK) in response to IL18.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Accessory protein-like; AcPL; CD218 antigen-like family member B; CD218b; CDw218b; cluster of differentiation w218b; IL-18 RA; IL-18 receptor accessory protein; IL-18 receptor beta; IL-18R-beta; IL-1R accessory protein-like; IL-1R-7; IL-1RAcPL; Interleukin-1 receptor 7; Interleukin-18 receptor accessory protein; Interleukin-18 receptor accessory protein-like; Interleukin-18 receptor beta

View more View less

Gene Aliases: ACPL; CD218b; CDw218b; IL-18R-beta; IL-18RAcP; IL-18Rbeta; IL-1R-7; IL-1R7; IL-1RAcPL; IL18RAP; IL18RB; IL1R7

View more View less

UniProt ID: (Human) O95256

View more View less

Entrez Gene ID: (Human) 8807

View more View less

Function(s)
receptor activity
Process(es)
inflammatory response immune response cell surface receptor signaling pathway cellular response to hydrogen peroxide cellular response to cytokine stimulus
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0