Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • IL10RB Antibodies

Invitrogen

IL10RB Monoclonal Antibody (90220)

View all (24) IL10RB antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite IL10RB Monoclonal Antibody (90220)

  • Antibody Testing Data (2)
IL10RB Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
IL10RB Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

IL10RB Antibody (MA5-23848) in Flow

Flow cytometry of IL10RB in Human PBMC monocytes. Samples were incubated in IL10RB monoclonal antibody (Product # MA5-23848) or isotype control antibody followed by PE-conjugated Anti-Mouse IgG Secondary Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

IL10RB Antibody in Flow Cytometry (Flow)
IL10RB Antibody in Neutralization (Neu)
IL10RB Monoclonal Antibody (90220)

Product Details

MA5-23848

Applications
Tested Dilution
Publications

Western Blot (WB)

1 µg/mL
-

Flow Cytometry (Flow)

0.25 µg per million cells
-

Neutralization (Neu)

0.5-2 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

90220

Immunogen

Mouse myeloma cell line NS0-derived recombinant human IL-10 R beta Met20-Ser220
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

0.5 mg/mL

Purification

Protein A/G

Storage buffer

PBS with 5% trehalose

Contains

No Preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2608053

Product Specific Information

In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-10 R alpha, rhIFN-gamma RI, and rhIFN-gamma R2 is observed.

Reconstitute at 0.5 mg/mL in sterile PBS.

Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

Target Information

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CDw210b; CRF2-4; cytokine receptor class-II CRF2-4; Cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; IL-10RB; inte; interleukin 10 receptor, beta; Interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta

View more View less

Gene Aliases: CDW210B; CRF2-4; CRFB4; D21S58; D21S66; IL-10R2; IL10RB

View more View less

UniProt ID: (Human) Q08334

View more View less

Entrez Gene ID: (Human) 3588

View more View less

Function(s)
receptor activity interleukin-10 receptor activity protein binding
Process(es)
inflammatory response immune response signal transduction cytokine-mediated signaling pathway defense response to virus
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0