Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • IL-5 Antibodies

Invitrogen

IL-5 Monoclonal Antibody (TRFK5), eFluor™ 450, eBioscience™

3 Published Figures
2 References
View all (45) IL-5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite IL-5 Monoclonal Antibody (TRFK5), eFluor™ 450, eBioscience™

  • Antibody Testing Data (1)
  • Published Figures (3)
IL-5 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
IL-5 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

IL-5 Antibody (48-7052-82) in Flow

Normal human peripheral blood cells were polarized under Th2 conditions and then unstimulated (left) or stimulated with the Cell Stimulation Cocktail (plus protein transport inhibitors) (Product # 00-4975-93) (right). Cells were then stained intracellularly, using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol, with CD4 Monoclonal Antibody, APC (Product # 17-0049-42) and IL-5 Monoclonal Antibody, eFluor 450. Total viable cells were used for analysis, as determined by Fixable Viability Dye eFluo... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

IL-5 Antibody in Flow Cytometry (Flow)
IL-5 Antibody in ELISA (ELISA)
IL-5 Antibody in ELISA (ELISA)
IL-5 Antibody in Neutralization (Neu)

Product Details

48-7052-82

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

0.25 µg/test
- -

ELISA (ELISA)

-
View 1 publication 1 publication

Neutralization (Neu)

-
View 1 publication 1 publication
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rat / IgG1

Recommended Isotype Control

Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 450, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

TRFK5

Conjugate

eFluor™ 450 eFluor™ 450 eFluor™ 450
  • Unconjugated
  • Functional Grade
  • PE 
  • Request custom conjugation

Excitation/Emission Max

405/445 nm View spectra spectra

Form

Liquid

Concentration

0.2 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2802295

Product Specific Information

Description: The TRFK5 antibody reacts with mouse and human interleukin-5 (IL-5). The TRFK5 antibody is a neutralizing antibody. mouse IL-5 is a disulfide-linked homodimer, containing two 113 amino acid peptides; these resolve as 32-34 kDa bands in SDS-PAGE. IL-5 is produced by T cells and has been known as eosinophil-differentiating factor (EDF), B cell growth factor II (BCGFII), and T cell-replacing factor (TRF). IL-5 induces eosinophil differentiation and promotes eosinophil survival and activation. In mice, IL-5 has been shown to stimulate B cell proliferation and antibody production.

Applications Reported: This TRFK5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

Applications Tested: This TRFK5 antibody has been tested by intracellular staining followed by flow cytometric analysis of Th2-polarized and stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to "Staining Intracellular Antigens for Flow Cytometry, Protocol A: Two step protocol for intracellular (cytoplasmic) proteins" located at www.thermofisher.com/flowprotocols . This may be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

eFluor 450 is an alternative to Pacific Blue. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome.

Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.

Target Information

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

Bioinformatics

Protein Aliases: B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; Cytotoxic T-lymphocyte inducer; Eosinophil differentiation factor; IL-5; ILN; Interleukin; Interleukin-5; Interleukin5; T-cell replacing factor; TRF

View more View less

Gene Aliases: EDF; IL-5; IL5; TRF

View more View less

UniProt ID: (Human) P05113, (Mouse) P04401

View more View less

Entrez Gene ID: (Human) 3567, (Mouse) 16191

View more View less

Function(s)
protein tyrosine kinase activity Ras guanyl-nucleotide exchange factor activity cytokine activity interleukin-5 receptor binding protein binding growth factor activity
Process(es)
immune response positive regulation of cell proliferation cytokine-mediated signaling pathway positive regulation of B cell proliferation positive regulation of eosinophil differentiation positive regulation of transcription, DNA-templated positive regulation of JAK-STAT cascade positive regulation of peptidyl-tyrosine phosphorylation positive regulation of immunoglobulin secretion positive regulation of sequence-specific DNA binding transcription factor activity positive regulation of podosome assembly MAPK cascade inflammatory response peptidyl-tyrosine phosphorylation positive regulation of GTPase activity
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0