Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • IL-33 Antibodies

Invitrogen

IL-33 Recombinant Rabbit Monoclonal Antibody (002)

View all (36) IL-33 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite IL-33 Recombinant Rabbit Monoclonal Antibody (002)

  • Antibody Testing Data (1)
IL-33 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
IL-33 Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

IL-33 Antibody (MA5-29362) in Flow

Flow cytometric analysis of Purified anti-Human IL-33 on HUVEC cells. HUVEC cells were treated according to manufacturer’s manual, and stained with IL-33 Recombinant Rabbit Monoclonal Antibody (2) (Product # MA5-29362) (Bold line hisgram), followed by a FITC-conjugated Secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

IL-33 Antibody in Flow Cytometry (Flow)
IL-33 Recombinant Rabbit Monoclonal Antibody (002)

Product Details

MA5-29362

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

1:25-1:100
-

ELISA (ELISA)

1:5,000-1:10,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

002

Immunogen

Recombinant Human IL-33 protein (Ser112-Thr270)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated
  • PE 
  • Request custom conjugation

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2785239

Product Specific Information

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This antibody has specificity for Human IL-33.

Target Information

IL-33 (Interleukin-33) is a 270 amino acid, highly divergent protein belonging to the IL-1 family with an IL-1-like C-terminal domain. IL-33 is a dual function protein that may function both as a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. IL-33 binds to and signals through IL1RL1/ST2 and its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6. IL-33 activates NF-kappaB and MAP kinases, and drives production of TH2-associated cytokines from in vitro polarized TH2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs. IL-33 is proteolytically converted to a mature form by CASP1 and is highly expressed in high endothelial venules found in tonsils, Peyer's patches and mesenteric lymph nodes and is almost undetectable in placenta. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine. Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. Despite its predicted molecular weight, IL-33 will often run at higher molecular weight in SDS-PAGE. Studies have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine, and an intracellular nuclear factor with transcriptional regulatory properties.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: DVS27-related protein; growth stimulation-expressed; H-IL-33; homolog of mouse growth stimulation-expressed; IL-1F11; IL-33; ILN; Interleukin; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin-1 family member 11; Interleukin-1 receptor-like 1; Interleukin-33; Interleukin33; NF-HEV; nuclear factor for high endothelial venules; Nuclear factor from high endothelial venules; Protein ST2; RP11-575C20.2

View more View less

Gene Aliases: C9orf26; DER4; DVS27; FIT-1; IL1F11; IL1RL1; IL33; IL33R; NF-HEV; NFEHEV; NFHEV; ST2; ST2L; ST2V; T1

View more View less

UniProt ID: (Human) Q01638, (Human) O95760

View more View less

Entrez Gene ID: (Human) 9173, (Human) 90865

View more View less

Function(s)
interleukin-33 binding interleukin-33 receptor activity cytokine receptor activity interleukin-1 receptor activity receptor signaling protein activity protein binding cytokine activity
Process(es)
negative regulation of leukocyte migration negative regulation of T-helper 1 type immune response positive regulation of type 2 immune response transcription, DNA-templated positive regulation of proteasomal ubiquitin-dependent protein catabolic process negative regulation of interferon-gamma production positive regulation of interleukin-13 production positive regulation of interleukin-4 production positive regulation of interleukin-5 production positive regulation of interleukin-6 production type 2 immune response positive regulation of macrophage activation positive regulation of transcription from RNA polymerase II promoter positive regulation of inflammatory response positive regulation of immunoglobulin secretion negative regulation of immunoglobulin secretion defense response to virus positive regulation of chemokine secretion extrinsic apoptotic signaling pathway immune response signal transduction negative regulation of cell proliferation interleukin-33-mediated signaling pathway negative regulation of I-kappaB kinase/NF-kappaB signaling cellular response to peptide
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0