Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • H-Ras Antibodies

OriGene

HRAS Monoclonal Antibody (OTI1H3), TrueMAB™

View all (43) H-Ras antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite HRAS Monoclonal Antibody (OTI1H3), TrueMAB™

  • Antibody Testing Data (2)
HRAS Antibody in Western Blot (WB)
Group 53 Created with Sketch.
HRAS Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

HRAS Antibody (CF505674) in WB

Western blot analysis of extracts (35 µg) from 9 different cell lines by using anti-HRAS monoclonal antibody (HepG2: human; HeLa: human; SVT2: mouse; A549: human; COS7: monkey; Jurkat: human; MDCK: canine; PC12: rat; MCF7: human). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

HRAS Antibody in Western Blot (WB)
HRAS Antibody in Western Blot (WB)
HRAS Monoclonal Antibody (OTI1H3), TrueMAB™

Product Details

CF505674

Applications
Tested Dilution
Publications

Western Blot (WB)

1:200-1:4000
-
Product Specifications

Species Reactivity

Dog, Human, Mouse, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI1H3

Immunogen

Full length human recombinant protein of human HRAS produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

Monomeric G protein Ras functions as a molecular switch linking receptor and nonreceptor tyrosine kinase activation to a number of downstream signaling pathways leading to distinct cytoplasmic or nuclear events. Each mammalian cell contains three Ras proto-oncogene coding for closely related Ras proteins: H-, K-, N-Ras. Oncogenic mutations in Ras genes are present in ~30% of all human cancers. Constitutive activation of Ras due to mutations or overexpression stimulates proliferation and inhibition of apoptosis. K-Ras mutations are common in pancreatic, colorectal and nonsmall-cell lung carcinomas; H-Ras mutations are common in bladder, kidney and thyroid carcinomas; N-Ras mutations are found in melanoma, hematological malignancies and hepatocellular carcinaoma.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: c-H-ras; c-Ha-ras p21 protein; c-Ha-ras transgene; c-has/bas p21 protein; c-ras-Ki-2 activated oncogene; GTP- and GDP-binding peptide B; GTPase HRas; GTPase Hras1; H-ras 1 protein; H-Ras-1; Ha-Ras; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene 1; p19 H-RasIDX protein; p21ras; Ras family small GTP binding protein H-Ras; ras p21; transformation gene: oncogene HAMSV; Transforming protein p21; v-Ha-ras Harvey rat sarcoma viral oncogene homolog

View more View less

Gene Aliases: C-BAS/HAS; C-H-RAS; c-Ha-ras; C-HA-RAS1; c-rasHa; CTLO; H-RAS; H-RASIDX; Ha-ras; HAMSV; Harvey-ras; HRAS; Hras-1; HRAS1; Kras2; p21ras; ras; RASH1

View more View less

UniProt ID: (Human) P01112, (Mouse) Q61411, (Rat) P20171

View more View less

Entrez Gene ID: (Dog) 403735, (Human) 3265, (Mouse) 15461, (Rat) 293621

View more View less

Function(s)
GTPase activity protein binding GTP binding protein C-terminus binding nucleotide binding
Process(es)
positive regulation of protein phosphorylation endocytosis apoptotic process cell cycle arrest mitotic cell cycle checkpoint signal transduction small GTPase mediated signal transduction Ras protein signal transduction cell aging cell proliferation positive regulation of cell proliferation negative regulation of cell proliferation positive regulation of gene expression negative regulation of gene expression positive regulation of cell migration negative regulation of GTPase activity positive regulation of MAP kinase activity positive regulation of MAPK cascade negative regulation of neuron apoptotic process positive regulation of GTPase activity positive regulation of DNA replication positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade positive regulation of Ras protein signal transduction regulation of long-term neuronal synaptic plasticity positive regulation of epithelial cell proliferation protein heterooligomerization positive regulation of ERK1 and ERK2 cascade positive regulation of wound healing cellular senescence intrinsic apoptotic signaling pathway positive regulation of ruffle assembly positive regulation of actin cytoskeleton reorganization positive regulation of miRNA metabolic process visual learning actin cytoskeleton organization regulation of synaptic transmission, GABAergic positive regulation of Rac protein signal transduction social behavior negative regulation of cell differentiation striated muscle cell differentiation epithelial tube branching involved in lung morphogenesis MAPK cascade stimulatory C-type lectin receptor signaling pathway chemotaxis cell surface receptor signaling pathway epidermal growth factor receptor signaling pathway axon guidance organ morphogenesis positive regulation of interferon-gamma production Fc-epsilon receptor signaling pathway ERBB2 signaling pathway T-helper 1 type immune response defense response to protozoan ephrin receptor signaling pathway T cell receptor signaling pathway leukocyte migration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0