Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • HO-1 Antibodies

Invitrogen

HO-1 Recombinant Rabbit Monoclonal Antibody (HL1780)

View all (38) HO-1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite HO-1 Recombinant Rabbit Monoclonal Antibody (HL1780)

  • Antibody Testing Data (3)
HO-1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
HO-1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

HO-1 Antibody (MA5-47178) in WB

Western blot analysis of HO-1 in wild-type (WT) and HMOX1 knockout (KO) HeLa cell extracts (30 µg) separated by 12% SDS-PAGE. Membrane was blotted with HO-1 Monoclonal antibody (Product # MA5-47178) using a dilution of 1:2,000 followed by HRP-conjugated anti-rabbit IgG secondary antibody. The signal was developed with Trident ECL plus-Enhanced. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

HO-1 Antibody in Western Blot (WB)
HO-1 Antibody in Western Blot (WB)
HO-1 Antibody in Western Blot (WB)

Product Details

MA5-47178

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:3,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

HL1780

Immunogen

Recombinant fragment of human Heme Oxygenase 1

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2938250

Product Specific Information

Store as concentrated solution.

Centrifuge briefly prior to opening vial.

Target Information

Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: heat shock protein, 32-kD; heme orygenase 1; heme oxygenase (decycling) 1; Heme oxygenase 1; HO-1

View more View less

Gene Aliases: bK286B10; HMOX1; HMOX1D; HO; HO-1; HO1; HSP32

View more View less

UniProt ID: (Human) P09601

View more View less

Entrez Gene ID: (Human) 3162

View more View less

Function(s)
heme oxygenase (decyclizing) activity phospholipase D activity signal transducer activity protein binding enzyme binding heme binding protein homodimerization activity metal ion binding
Process(es)
angiogenesis endothelial cell proliferation wound healing involved in inflammatory response negative regulation of leukocyte migration heme oxidation cellular iron ion homeostasis response to oxidative stress small GTPase mediated signal transduction excretion regulation of blood pressure cell death intrinsic apoptotic signaling pathway in response to DNA damage negative regulation of muscle cell apoptotic process smooth muscle hyperplasia cellular response to nutrient negative regulation of mast cell cytokine production erythrocyte homeostasis low-density lipoprotein particle clearance regulation of transcription from RNA polymerase II promoter in response to iron cellular response to heat response to nicotine intracellular signal transduction heme catabolic process response to hydrogen peroxide positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of mast cell degranulation negative regulation of DNA binding negative regulation of sequence-specific DNA binding transcription factor activity negative regulation of neuron apoptotic process regulation of transcription from RNA polymerase II promoter in response to oxidative stress response to estrogen positive regulation of chemokine biosynthetic process regulation of angiogenesis positive regulation of angiogenesis positive regulation of vasodilation positive regulation of smooth muscle cell proliferation negative regulation of smooth muscle cell proliferation regulation of sequence-specific DNA binding transcription factor activity protein homooligomerization iron ion homeostasis cellular response to arsenic-containing substance cellular response to cadmium ion cellular response to hypoxia negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0