Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Secondary Antibodies ›
  • Goat anti-Mouse IgG Secondary Antibodies

Invitrogen

Goat anti-Mouse IgG (H+L) Secondary Antibody, FITC

View all (240) Goat anti-Mouse IgG antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Create Citation
Questions & Answers
Datasheet
Create Citation
Questions & Answers

Cite Goat anti-Mouse IgG (H+L) Secondary Antibody, FITC

Product Details

A16067

Applications
Tested Dilution

Immunocytochemistry (ICC/IF)

1:500-1:20,000

Flow Cytometry (Flow)

1:500-1:20,000
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Goat / IgG

Class

Polyclonal

Type

Secondary Antibody

Immunogen

Gamma Immunoglobins Heavy and Light chains

Conjugate

FITC FITC FITC
  • Agarose
  • Alexa Fluor 488 
  • Alexa Fluor 488-10 nm colloidal gold
  • Alexa Fluor 555 
  • Alexa Fluor 647 
  • Alexa Fluor 680 
  • Alexa Fluor 790 
  • Alexa Fluor Plus 405 
  • Alexa Fluor Plus 488 
  • Alexa Fluor Plus 647 
  • AP
  • bGal
  • Biotin 
  • DyLight 350
  • DyLight 405
  • DyLight 488
  • DyLight 550
  • DyLight 594
  • DyLight 633
  • DyLight 650
  • DyLight 680
  • DyLight 800 4X PEG
  • HRP
  • PE 
  • Rhodamine
  • TRITC
  • WesternDot 625
  • Request custom conjugation

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

2.5 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.8, with 1% BSA

Contains

0.05% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2534740

Target

IgG

Antibody Form

Whole Antibody

Product Specific Information

The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).

Target Information

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

View product citations for antibody A16067 on CiteAb

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0