Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Glypican 3 Antibodies

NeoBiotechnologies

Glypican-3 (GPC3) Recombinant Mouse Monoclonal Antibody (rGPC3, 863)

View all (58) Glypican 3 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Glypican-3 (GPC3) Recombinant Mouse Monoclonal Antibody (rGPC3, 863)

  • Antibody Testing Data (5)
Glypican-3 (GPC3) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Glypican-3 (GPC3) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Glypican-3 (GPC3) Antibody (2719-MSM6-P1) in ICC/IF

Immunocytochemistry/Immunofluorescence analysis of MeOH-fixed HepG2 cells using Glypican-3 (GPC3) Recombinant Monoclonal Antibody (Product # 2719-MSM6-P1) followed by goat anti-Mouse IgG-CF488 (Green). The nuclear counterstain is Reddot (Red).. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Glypican-3 (GPC3) Antibody in Immunocytochemistry (ICC/IF)
Glypican-3 (GPC3) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Glypican-3 (GPC3) Antibody in Flow Cytometry (Flow)
Glypican-3 (GPC3) Antibody in Peptide array (ARRAY)
Glypican-3 (GPC3) Antibody in SDS-PAGE (SDS-PAGE)
Glypican-3 (GPC3) Recombinant Mouse Monoclonal Antibody (rGPC3, 863)

Product Details

2719-MSM6-P1

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1-2 µg/mL
-

Immunocytochemistry (ICC/IF)

1-2 µg/mL
-

Flow Cytometry (Flow)

1-2 µg/10^6 cells
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human, Rat

Host/Isotype

Mouse / IgG1, kappa

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

rGPC3, 863

Immunogen

Recombinant full-length human GPC3 protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Immunohistochemistry (PFA fixed): incubate antibody for 30 min RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.

Target Information

GPC3 is a cell surface proteoglycan that bears heparan sulfate. This protein may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol (GPI) linkage. These proteins may play a role in the control of cell division, growth regulation, and tumor predisposition. Deletion mutations in GPC3 are the cause of Simpson-Golabi-Behmel syndrome (SGBS), also known as Simpson dysmorphia syndrome (SDYS). SGBS is a condition characterized by pre- and postnatal overgrowth (gigantism) with visceral and skeletal anomalies.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: defective in Simpson-Golabi-Behmel overgrowth syndrome; glypican proteoglycan 3; Glypican-3; GTR2-2; GTR22; heparan sulphate proteoglycan; Intestinal protein OCI-5; MXR7; proteoglycan GPC3; secreted glypican-3

View more View less

Gene Aliases: DGSX; GPC3; GTR2-2; MXR7; OCI-5; OCI5; SDYS; SGB; SGBS; SGBS1

View more View less

UniProt ID: (Human) P51654, (Rat) P13265

View more View less

Entrez Gene ID: (Human) 2719, (Rat) 25236

View more View less

Function(s)
protein binding heparan sulfate proteoglycan binding peptidyl-dipeptidase inhibitor activity molecular_function
Process(es)
branching involved in ureteric bud morphogenesis kidney development biological_process negative regulation of cell proliferation organ morphogenesis anterior/posterior axis specification body morphogenesis negative regulation of peptidase activity bone mineralization osteoclast differentiation lung development positive regulation of BMP signaling pathway embryonic hindlimb morphogenesis regulation of growth positive regulation of protein catabolic process positive regulation of endocytosis negative regulation of smoothened signaling pathway positive regulation of smoothened signaling pathway negative regulation of growth positive regulation of glucose import negative regulation of epithelial cell proliferation coronary vasculature development cell proliferation involved in kidney development mesenchymal cell proliferation involved in ureteric bud development mesonephric duct morphogenesis cell proliferation involved in metanephros development negative regulation of canonical Wnt signaling pathway positive regulation of Wnt signaling pathway, planar cell polarity pathway retinoid metabolic process glycosaminoglycan biosynthetic process glycosaminoglycan catabolic process anatomical structure morphogenesis glycosaminoglycan metabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-74848bcbc5-fbw66:80/100.66.128.25:80.
git-commit: ee75265227445a99bc079a1fecc2a603f2c9394f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0