Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GPNMB Antibodies

Proteintech

GPNMB Monoclonal Antibody (2B10B8)

1 Reference
View all (38) GPNMB antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GPNMB Monoclonal Antibody (2B10B8)

  • Antibody Testing Data (4)
GPNMB Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
GPNMB Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GPNMB Antibody (66926-1-IG) in ICC/IF

Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using 66926-1-Ig (GPNMB antibody) at dilution of 1:100 and CoraLite488-Conjugated AffiniPure Goat Anti-Mouse IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GPNMB Antibody in Immunocytochemistry (ICC/IF)
GPNMB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GPNMB Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GPNMB Antibody in Western Blot (WB)

Product Details

66926-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:10,000
-

Immunohistochemistry (IHC)

-
View 1 publication 1 publication

Immunohistochemistry (Paraffin) (IHC (P))

1:1,000
-

Immunocytochemistry (ICC/IF)

1:100
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

2B10B8

Immunogen

GPNMB Fusion Protein Ag26747

Conjugate

Unconjugated Unconjugated Unconjugated
  • CoraLite 594
  • CoraLite Plus 488
  • Request custom conjugation

Form

Liquid

Concentration

1000 µg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Aliquoting is unnecessary for -20°C storage.

Target Information

The protein encoded by this gene is a type I transmembrane glycoprotein which shows homology to the pMEL17 precursor, a melanocyte-specific protein. GPNMB shows expression in the lowly metastatic human melanoma cell lines and xenografts but does not show expression in the highly metastatic cell lines. GPNMB may be involved in growth delay and reduction of metastatic potential. Two transcript variants encoding different isoforms have been found for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: DC-HIL; dendritic cell-associated heparin sulfate proteoglycan-dependent integrin ligand; Dendritic cell-associated transmembrane protein; glycoprotein (transmembrane) nmb; glycoprotein nmb-like protein; glycoprotein nonmetastatic melanoma protein B; hematopoietic growth factor inducible neurokinin-1; Hematopoietic growth factor inducible neurokinin-1 type; iris pigment dispersion; Osteoactivin; transmembrane glycoprotein HGFIN; Transmembrane glycoprotein NMB

View more View less

Gene Aliases: DC-HIL; Dchil; GPNMB; HGFIN; ipd; NMB; UNQ1725/PRO9925

View more View less

UniProt ID: (Human) Q14956, (Mouse) Q99P91

View more View less

Entrez Gene ID: (Human) 10457, (Rat) 113955, (Mouse) 93695

View more View less

Function(s)
integrin binding heparin binding
Process(es)
osteoblast differentiation cell adhesion bone mineralization negative regulation of cell proliferation negative regulation of tumor necrosis factor production
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0