Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GBE1 Antibodies

Abnova

GBE1 Polyclonal Antibody, MaxPab™

View all (26) GBE1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GBE1 Polyclonal Antibody, MaxPab™

  • Antibody Testing Data (1)
GBE1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
GBE1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GBE1 Antibody (H00002632-B01P) in WB

Western Blot analysis of GBE1 expression in transfected 293T cell line by GBE1 MaxPab polyclonal antibody. Lane 1: GBE1 transfected lysate(77.22 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GBE1 Antibody in Western Blot (WB)

Product Details

H00002632-B01P

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG

Class

Polyclonal

Type

Antibody

Immunogen

GBE1 (AAH12098.1, 1 a.a. approximately 702 a.a) full-length human protein.

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Product may be used with Western Blot (Transfected lysate).

The Isotype of this product is composed of an IgG Mixture.

Immunogen sequence: MAAPMTPAAR PEDYEAALNA ALADVPELAR LLEIDPYLKP YAVDFQRRYK QFSQILKNIG ENEGGIDKFS RGYESFGVHR CADGGLYCKE WAPGAEGVFL TGDFNGWNPF SYPYKKLDYG KWELYIPPKQ NKSVLVPHGS KLKVVITSKS GEILYRISPW AKYVVREGDN VNYDWIHWDP EHSYEFKHSR PKKPRSLRIY ESHVGISSHE GKVASYKHFT CNVLPRIKGL GYNCIQLMAI MEHAYYASFG YQITSFFAAS SRYGSPEELQ ELVDTAHSMG IIVLLDVVHS HASKNSADGL NMFDGTDSCY FHSGPRGTHD LWDSRLFAYS SWEVLRFLLS NIRWWLEEYR FDGFRFDGVT SMLYHHHGVG QGFSGDYSEY FGLQVDEDAL TYLMLANHLV HTLCPDSITI AEDVSGMPAL CSPISQGGGG FDYRLAMAIP DKWIQLLKEF KDEDWNMGDI VYTLTNRRYL EKCIAYAESH DQALVGDKSL AFWLMDAEMY TNMSVLTPFT PVIDRGIQLH KMIRLITHGL GGEGYLNFMG NEFGHPEWLD FPRKGNNESY HYARRQFHLT DDDLLRYKFL NNFDRDMNRL EERYGWLAAP QAYVSEKHEG NKIIAFERAG LLFIFNFHPS KSYTDYRVGT ALPGKFKIVL DSDAAEYGGH QRLDHSTDFF SEAFEHNGRP YSLLVYIPSR VALILQNVDL PN

Target Information

GBE1 is a glycogen branching enzyme that catalyzes the transfer of alpha-1,4-linked glucosyl units from the outer end of a glycogen chain to an alpha-1,6 position on the same or a neighboring glycogen chain. Branching of the chains is essential to increase the solubility of the glycogen molecule and, consequently, in reducing the osmotic pressure within cells. Highest level of this enzyme are found in liver and muscle. Mutations in this gene are associated with glycogen storage disease IV (also known as Andersen's disease).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 1,4-alpha-glucan-branching enzyme; amylo-(1,4 to 1,6) transglucosidase; amylo-(1,4 to 1,6) transglycosylase; Brancher enzyme; glycogen branching enzyme; Glycogen-branching enzyme

View more View less

Gene Aliases: APBD; GBE; GBE1; GSD4

View more View less

UniProt ID: (Human) Q04446

View more View less

Entrez Gene ID: (Human) 2632

View more View less

Function(s)
1,4-alpha-glucan branching enzyme activity hydrolase activity, hydrolyzing O-glycosyl compounds cation binding
Process(es)
glycogen metabolic process glycogen biosynthetic process generation of precursor metabolites and energy
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0