Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GAPDHS Antibodies

Proteintech

GAPDHS Polyclonal Antibody

14 References
View all (15) GAPDHS antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GAPDHS Polyclonal Antibody

  • Antibody Testing Data (2)
GAPDHS Antibody in Western Blot (WB)
Group 53 Created with Sketch.
GAPDHS Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GAPDHS Antibody (13937-1-AP) in WB

Mouse testis tissue were subjected to SDS PAGE followed by western blot with 13937-1-AP (GAPDHS antibody) at dilution of 1:500 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GAPDHS Antibody in Western Blot (WB)
GAPDHS Antibody in Western Blot (WB)
GAPDHS Polyclonal Antibody

Product Details

13937-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
View 14 publications 14 publications
Product Specifications

Species Reactivity

Human, Mouse, Rat

Published species

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

GAPDHS Fusion Protein Ag4988 (58-408 aa encoded by BC036373)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.43 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: LPPHPATPP PKMVSVAREL TVGINGFGRI GRLVLRACME KGVKVVAVND PFIDPEYMVY MFKYDSTHGR YKGSVEFRNG QLVVDNHEIS VYQCKEPKQI PWRAVGSPYV VESTGVYLSI QAASDHISAG AQRVVISAPS PDAPMFVMGV NENDYNPGSM NIVSNASCTT NCLAPLAKVI HERFGIVEGL MTTVHSYTAT QKTVDGPSRK AWRDGRGAHQ NIIPASTGAA KAVTKVIPEL KGKLTGMAFR VPTPDVSVVD LTCRLAQPAP YSAIKEAVKA AAKGPMAGIL AYTEDEVVST DFLGDTHSSI FDAKAGIALN DNFVKLISWY DNEYGYSHRV VDLLRYMFSR DK (58-408 aa encoded by BC036373)

Target Information

Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH) is a metabolic enzyme responsible for catalyzing one step in the glycolytic pathway, the reversible oxidative phosphorylation of glyceraldehyde 3-phosphate. Because GAPDH is a protein expressed in large amounts and which is required at all times for important "house keeping" functions, levels of GAPDH mRNA are often measured and used as standards in studies of mRNA expression. Increasingly, scientists are making use of specific antibodies to GAPDH in comparable studies of levels of protein expression. GAPDH is reported to bind to a variety of other proteins, including the amyloid precursor protein, mutations in which cause some forms of Alzheimer's disease, and the polyglutamine tracts of Huntingtin, the protein product aberrant forms of which are causative of Huntington's disease. Associations with actin and tubulin have also been reported. The protein may also have a role in the regulation of apoptosis, and interestingly migrates from the cytoplasm into the nucleus when cells become apoptotic.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: epididymis secretory protein Li 278; GAPDH S; GAPDH-2; glyceraldehyde-3-phosphate dehydrogenase type 2; Glyceraldehyde-3-phosphate dehydrogenase, testis-specific; Spermatogenic cell-specific glyceraldehyde 3-phosphate dehydrogenase 2; Spermatogenic glyceraldehyde-3-phosphate dehydrogenase

View more View less

Gene Aliases: Gapd-s; GAPD2; GAPDH-2; GAPDH2; GAPDHS; GAPDS; HEL-S-278; HSD-35; HSD35

View more View less

UniProt ID: (Human) O14556, (Rat) Q9ESV6, (Mouse) Q64467

View more View less

Entrez Gene ID: (Human) 26330, (Rat) 66020, (Mouse) 14447

View more View less

Function(s)
glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity protein binding NADP binding NAD binding molecular_function oxidoreductase activity oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
Process(es)
gluconeogenesis spermatid development sperm motility positive regulation of glycolytic process canonical glycolysis glucose metabolic process glycolytic process oxidation-reduction process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0