Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GABRA1 Antibodies

Proteintech

GABRA1 Polyclonal Antibody

29 References
View all (26) GABRA1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GABRA1 Polyclonal Antibody

  • Antibody Testing Data (7)
GABRA1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
GABRA1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 7

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GABRA1 Antibody (12410-1-AP) in WB

Mouse brain tissue were subjected to SDS PAGE followed by western blot with (Product # 12410-1-AP) GABRA1 antibody at dilution of 1:2,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GABRA1 Antibody in Western Blot (WB)
GABRA1 Antibody in Western Blot (WB)
GABRA1 Antibody in Western Blot (WB)
GABRA1 Antibody in Western Blot (WB)
GABRA1 Antibody in Western Blot (WB)
GABRA1 Antibody in Flow Cytometry (Flow)
GABRA1 Antibody in Immunoprecipitation (IP)
GABRA1 Polyclonal Antibody

Product Details

12410-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
View 21 publications 21 publications

Immunocytochemistry (ICC/IF)

Assay-dependent
View 4 publications 4 publications

Flow Cytometry (Flow)

0.2 µg/1x10^6 cells
View 1 publication 1 publication

Immunoprecipitation (IP)

0.5-4.0 µg
View 1 publication 1 publication

Not applicable (N/A)

-
View 2 publications 2 publications
Product Specifications

Species Reactivity

Human, Mouse, Rat

Published species

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

GABRA1 Fusion Protein Ag3117 (21-280 aa encoded by BC030696)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.23 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: LTGRSYGQP SLQDELKDNT TVFTRILDRL LDGYDNRLRP GLGERVTEVK TDIFVTSFGP VSDHDMEYTI DVFFRQSWKD ERLKFKGPMT VLRLNNLMAS KIWTPDTFFH NGKKSVAHNM TMPNKLLRIT EDGTLLYTMR LTVRAECPMH LEDFPMDAHA CPLKFGSYAY TRAEVVYEWT REPARSVVVA EDGSRLNQYD LLGQTVDSGI VQSSTGEYVV MTTHFHLKRK IGYFVIQTYL PCIMTVILSQ VSFWLNRESV P (21-280 aa encoded by BC030696)

Target Information

Gamma -aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABAA Receptor (GABAA-R) subtype. GABAA-Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABAA-R is a multimeric subunit complex. To date aix alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for alpha- and beta-subunits results in the expression of functional GABAA-R sensitive to GABA. However, coexpression of a gamma-subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha-subunits of the receptor. Lastly, phosphorylation of beta-subunits of the receptor has been shown to modulate GABAA-R function.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: GABA(A) receptor subunit alpha-1; GABA(A) receptor, alpha 1; GABAA alpha 1; GABAAR alpha1; GABAAR subunit alpha-1; GABRA 1; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 1; gamma-aminobutyric acid (GABA) A receptor, alpha 1; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 1; gamma-aminobutyric acid A receptor, alpha 1; Gamma-aminobutyric acid receptor subunit alpha-1

View more View less

Gene Aliases: ECA4; EIEE19; EJM; EJM5; Gabra-1; GABRA1

View more View less

UniProt ID: (Human) P14867, (Rat) P62813, (Mouse) P62812

View more View less

Entrez Gene ID: (Human) 2554, (Rat) 29705, (Mouse) 14394

View more View less

Function(s)
GABA-A receptor activity drug binding GABA receptor activity GABA-gated chloride ion channel activity inhibitory extracellular ligand-gated ion channel activity chloride channel activity protein binding benzodiazepine receptor activity ion channel activity extracellular ligand-gated ion channel activity
Process(es)
transport gamma-aminobutyric acid signaling pathway ion transmembrane transport synaptic transmission, GABAergic cellular response to histamine chloride transmembrane transport chloride transport ion transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0