Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • FITC Antibodies

Invitrogen

FITC Chimeric Recombinant Mouse Monoclonal Antibody (4-4-20 (enhanced))

View all (65) FITC antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite FITC Chimeric Recombinant Mouse Monoclonal Antibody (4-4-20 (enhanced))

Product Details

MA5-51708

Applications
Tested Dilution
Publications

Functional Assay (Functional)

Assay-dependent
-
Product Specifications

Species Reactivity

Chemical

Host/Isotype

Mouse / IgG2a, kappa

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

4-4-20 (enhanced)

Immunogen

Fluorescein I-keyhole limpet hemocyanin (FlI-KLH).

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS

Contains

0.02% ProClin 300

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3250400

Product Specific Information

This antibody is specific for fluorescein I.

Target Information

FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs ultraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-8c6d775fd-xc6bz:80/100.66.129.86:80.
git-commit: 22e90aa5931c1aba9e24bf54669fab5290434a03
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0