Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • EOMES Antibodies

Invitrogen

EOMES Monoclonal Antibody (Dan11mag), PE, eBioscience™

26 References
View all (36) EOMES antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite EOMES Monoclonal Antibody (Dan11mag), PE, eBioscience™

  • Antibody Testing Data (1)
EOMES Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
EOMES Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

EOMES Antibody (12-4875-82) in Flow

Surface staining of C57BL/6 splenocytes with Anti-Mouse NK1-1 eFluor® 450 (Product # 48-5941-82) followed by intracellular staining with 0.25 µg of Rat IgG2a K Isotype Control PE (Product # 12-4321-80) (left) or 0.25 µg of Anti-Mouse EOMES PE (right) using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

EOMES Antibody in Flow Cytometry (Flow)

Product Details

12-4875-82

Applications
Tested Dilution
Publications

Immunohistochemistry (IHC)

-
View 6 publications 6 publications

Immunocytochemistry (ICC/IF)

-
View 1 publication 1 publication

Flow Cytometry (Flow)

0.5 µg/test
View 19 publications 19 publications
Product Specifications

Species Reactivity

Mouse

Published species

Mouse

Host/Isotype

Rat / IgG2a, kappa

Recommended Isotype Control

Rat IgG2a kappa Isotype Control (eBR2a), PE, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

Dan11mag

Conjugate

PE PE PE
  • Unconjugated
  • Alexa Fluor 488 
  • Alexa Fluor 660 
  • APC 
  • eFluor 450
  • eFluor 660
  • PE-Cyanine5
  • PE-Cyanine7
  • PE-eFluor 610
  • PerCP-eFluor 710
  • Request custom conjugation

Excitation/Emission Max

565/576 nm View spectra spectra

Form

Liquid

Concentration

0.2 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_1603275

Product Specific Information

Description: This Dan11mag antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R.

Applications Reported: This Dan11mag antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

Applications Tested: This Dan11mag antibody has been tested by intracellular flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please see BestProtocols® Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

Filtration: 0.2 µm post-manufacturing filtered.

Target Information

TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

Bioinformatics

Protein Aliases: Eomesodermin homolog; T-box brain protein 2; T-brain-2

View more View less

Gene Aliases: C77258; Eomes; TBR-2; Tbr2

View more View less

UniProt ID: (Mouse) O54839

View more View less

Entrez Gene ID: (Mouse) 13813

View more View less

Function(s)
RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II activating transcription factor binding transcriptional repressor activity, RNA polymerase II transcription factor binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding sequence-specific DNA binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter endoderm formation mesoderm formation endodermal cell fate specification blastocyst development trophectodermal cell differentiation CD8-positive, alpha-beta T cell differentiation involved in immune response immune system process transcription, DNA-templated regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter multicellular organism development gastrulation brain development cardioblast differentiation regulation of gene expression stem cell population maintenance olfactory bulb development cerebral cortex regionalization cerebral cortex neuron differentiation cell differentiation interferon-gamma production skeletal muscle cell differentiation positive regulation of cell differentiation regulation of neuron differentiation positive regulation of transcription from RNA polymerase II promoter regulation of neurogenesis cell differentiation involved in embryonic placenta development mesodermal to mesenchymal transition involved in gastrulation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0