Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cytokeratin 17 Antibodies

Progen

Cytokeratin 17 Monoclonal Antibody (Ks17.E3)

View all (53) Cytokeratin 17 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Cytokeratin 17 Monoclonal Antibody (Ks17.E3)

  • Antibody Testing Data (8)
Cytokeratin 17 Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.
Cytokeratin 17 Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.

FIGURE: 1 / 8

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Cytokeratin 17 Antibody (690036PROGEN) in IHC

Immunohistochemistry of rat bladder (courtesy of J.H, University Hospital Heidelberg). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Cytokeratin 17 Antibody in Immunohistochemistry (IHC)
Cytokeratin 17 Antibody in Immunohistochemistry (IHC)
Cytokeratin 17 Antibody in Immunohistochemistry (IHC)
Cytokeratin 17 Antibody in Immunohistochemistry (IHC)
Cytokeratin 17 Antibody in Immunohistochemistry (IHC)
Cytokeratin 17 Antibody in Immunohistochemistry (IHC)
Cytokeratin 17 Antibody in Immunohistochemistry (IHC)
Cytokeratin 17 Antibody in Western Blot (WB)
Cytokeratin 17 Monoclonal Antibody (Ks17.E3)

Product Details

690036PROGEN

Applications
Tested Dilution
Publications

Western Blot (WB)

1:100
-

Immunohistochemistry (IHC)

Assay-dependent
-

Immunohistochemistry (Paraffin) (IHC (P))

1:10-1:50
-

Immunohistochemistry (Frozen) (IHC (F))

1:10-1:50
-
Product Specifications

Species Reactivity

Human, Rat

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

Ks17.E3

Immunogen

Human Keratin K17 Of 46 Kda
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

50 µg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4, with 0.5% BSA

Contains

0.09% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Ks 17.E3 represents an excellent marker to distinguish myoepithelial cells (positive for K17) from luminal epithelium of various glands (mammary, sweat, salivary, bronchial, tracheal, laryngeal, esophageal) and benign from malignant forms of e.g. mammary gland tumors. In the epidermis suprabasal staining is found only in cornifying regions and in the outer root sheath of hair follicles.Tested reactivity on cultured cell lines: HeLa, A-431Tumors specifically detected: Benign tumors of mammary gland; most malignant forms of mammary tumors are negative for keratin K17.

Target Information

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9 - 7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 39.1; CK-17; CK17; Cytokeratin-17; hard keratin; K17; keratin 17, type I; Keratin, type I cytoskeletal 17; Keratin-17; krt17 {ECO:0000250|UniProtKB:Q04695}; Type I keratin Ka17

View more View less

Gene Aliases: 39.1; CK-17; K17; Ka17; KRT17; PC; PC2; PCHC1

View more View less

UniProt ID: (Human) Q04695, (Rat) Q6IFU8

View more View less

Entrez Gene ID: (Human) 3872, (Rat) 287702

View more View less

Function(s)
structural constituent of cytoskeleton protein binding MHC class II receptor activity MHC class II protein binding
Process(es)
signal transduction epidermis development positive regulation of cell growth hair follicle morphogenesis keratinization intermediate filament organization positive regulation of translation positive regulation of hair follicle development morphogenesis of an epithelium intermediate filament-based process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0