Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cytochrome C Antibodies

NeoBiotechnologies

Cytochrome C (Mitochondrial Marker) Monoclonal Antibody (7H8.2C12)

View all (67) Cytochrome C antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Cytochrome C (Mitochondrial Marker) Monoclonal Antibody (7H8.2C12)

  • Antibody Testing Data (3)
Cytochrome C (Mitochondrial Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Cytochrome C (Mitochondrial Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Cytochrome C (Mitochondrial Marker) Antibody (54205-MSM2-P1ABX) in IHC (P)

Formalin-fixed, paraffin-embedded human Pancreas stained with Cytochrome C Mouse Monoclonal Antibody (7H8.2C12). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Cytochrome C (Mitochondrial Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cytochrome C (Mitochondrial Marker) Antibody in Western Blot (WB)
Cytochrome C (Mitochondrial Marker) Antibody in SDS-PAGE (SDS-PAGE)

Product Details

54205-MSM2-P1ABX

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-
Product Specifications

Species Reactivity

Amphibian, Avian, Dog, Fruit fly, Horse, Human, Mouse, Rat

Host/Isotype

Mouse / IgG2b, kappa

Class

Monoclonal

Type

Antibody

Clone

7H8.2C12

Immunogen

Synthetic peptides corresponding to amino acid 1-80, 81-104 and 66-104 of pigeon cytochrome c

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Target Information

Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CG17903-PA; CG17903-PB; CRF; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; cytochrome c proximal; Cytochrome C, expressed in somatic tissues; Cytochrome c, somatic; Cytochrome c-2; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; hc1

View more View less

Gene Aliases: CG17903; CYC; CYCS; CYCSA; Cyct-c-p; Cyt C; cyt c-p; Cyt-c-p; Cyt-c2; cyt.c; CytC; CytC-2; Cytc-p; CYTC2; cytochrome c; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; HCS; pDMc01; THC4

View more View less

UniProt ID: (Dog) P00011, (Human) P99999, (Horse) P00004, (Mouse) P62897, (Rat) P62898, (Fruit fly) P84029

View more View less

Entrez Gene ID: (Dog) 475258, (Human) 54205, (Horse) 100053958, (Mouse) 13063, (Rat) 25309, (Fruit fly) 34996

View more View less

Function(s)
heme binding electron transporter, transferring electrons from CoQH2-cytochrome c reductase complex and cytochrome c oxidase complex activity metal ion binding protein serine/threonine phosphatase activity protein binding electron carrier activity iron ion binding lipid binding
Process(es)
mitochondrial electron transport, ubiquinol to cytochrome c mitochondrial electron transport, cytochrome c to oxygen apoptotic DNA fragmentation positive regulation of cysteine-type endopeptidase activity involved in apoptotic process response to reactive oxygen species protein dephosphorylation mitochondrion organization activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c cellular respiration intrinsic apoptotic signaling pathway apoptotic process hydrogen peroxide metabolic process ATP synthesis coupled electron transport oxidative phosphorylation response to ischemia response to oxidative stress negative regulation of hydrogen peroxide biosynthetic process glial cell apoptotic process positive regulation of cellular respiration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5f5c96ff6c-9qlkd:80/100.66.135.177:80.
git-commit: 945c3ce0b94337c0c8e9574aed9898c23d0d1109
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0