Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cyclin A2 Antibodies

NeoBiotechnologies

Cyclin A2 (S- and G2-phase Cyclin) Monoclonal Antibody (CCNA2/2333)

View all (31) Cyclin A2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Cyclin A2 (S- and G2-phase Cyclin) Monoclonal Antibody (CCNA2/2333)

  • Antibody Testing Data (3)
Cyclin A2 (S- and G2-phase Cyclin) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Cyclin A2 (S- and G2-phase Cyclin) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Cyclin A2 (S- and G2-phase Cyclin) Antibody (890-MSM3-P0) in IHC (P)

Formalin-fixed, paraffin-embedded human Endometrial Carcinoma stained with Cyclin A2 Mouse Monoclonal Antibody (CCNA2/2333). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Cyclin A2 (S- and G2-phase Cyclin) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cyclin A2 (S- and G2-phase Cyclin) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Cyclin A2 (S- and G2-phase Cyclin) Antibody in SDS-PAGE (SDS-PAGE)
Cyclin A2 (S- and G2-phase Cyclin) Monoclonal Antibody (CCNA2/2333)

Product Details

890-MSM3-P0

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

CCNA2/2333

Immunogen

Full length recombinant Cyclin A2 protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Wet ice

Target Information

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdks) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdks. These cyclin / cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S phase and passage through G2-phase.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Cyclin A; Cyclin-A; Cyclin-A2

View more View less

Gene Aliases: CCN1; CCNA; CCNA2

View more View less

UniProt ID: (Human) P20248

View more View less

Entrez Gene ID: (Human) 890

View more View less

Function(s)
protein binding protein kinase binding
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity mitotic nuclear division mitotic G2 DNA damage checkpoint Ras protein signal transduction viral process organ regeneration response to glucagon cellular response to platelet-derived growth factor stimulus cellular response to leptin stimulus positive regulation of transcription, DNA-templated positive regulation of fibroblast proliferation cell division cellular response to cocaine cellular response to luteinizing hormone stimulus cellular response to estradiol stimulus cellular response to hypoxia cellular response to nitric oxide cochlea development cellular response to insulin-like growth factor stimulus
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0