ELTI
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4 C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the product.
Species Reactivity: Cross Reactivity 2,4-DB 18% fenitrothion 13% Bezafibrate 11% Fenofibrate 6% Clofibrate 100% Clofibrinic acid 4% Chlorpyriphos 3% MCPA-Na 3% Mecoprop 2% 2,4,5-T 1% 2,4-D 1% 2,4-DP 1% Ciprofibrate 1% Gemfibrozil 1% MCPA 1% Pirimifos-ethyl 1% Diclofenac <0.1% Erythromycine <0.1% Propranolol <0.1%.
Application notes: Suggested concentration to use in ELISA: 1/15,000 from the delivered solution. Plates are coated with 400 ng/mL BSA-conjugated C6-alkylphenol. HRP-conjugated anti-rabbit IgG as a tracer 1/8,000.
Clofibrate is a drug used for the treatment of cardiovascular diseases as a lipid-lowering agent. It is found in the water environment in relatively high concentrations, especially in the neighbourhood of large cities and hospitals. It is rapidly degraded to clofibric acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support