Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Caspase 1 Antibodies

Invitrogen

Caspase 1 Polyclonal Antibody

View all (25) Caspase 1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Caspase 1 Polyclonal Antibody

  • Antibody Testing Data (16)
Caspase 1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Caspase 1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 16

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Caspase 1 Antibody (PA5-119001) in ICC/IF

Immunocytochemistry analysis of Caspase 1 in Human U2OS cells using Caspase 1 Polyclonal Antibody (Product # PA5-119001) at a dilution of 30 µg/mL, followed by secondary antibody staining with 2 µg/mL FITC-Linked Caprine Anti-Rabbit IgG Polyclonal Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Caspase 1 Antibody in Immunocytochemistry (ICC/IF)
Caspase 1 Antibody in Immunocytochemistry (ICC/IF)
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Caspase 1 Antibody in Western Blot (WB)
Caspase 1 Antibody in Western Blot (WB)
Caspase 1 Antibody in Western Blot (WB)
Caspase 1 Polyclonal Antibody

Product Details

PA5-119001

Applications
Tested Dilution
Publications

Western Blot (WB)

0.01-2 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

5-20 µg/mL
-

Immunocytochemistry (ICC/IF)

5-30 µg/mL
-
Product Specifications

Species Reactivity

Human, Pig

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant protein Caspase 1. The antigen corresponds to amino acid range 120-297 of the target protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5 mg/mL

Purification

Antigen affinity chromatography, Protein A

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2903501

Target Information

Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12 and can activate proinflammatory cytokines such as IL-1-b and IL-18. Caspase-1 was initially identified as an IL-1-b-converting enzyme; later experiments revealed it to be a mammalian homolog of the C. elegans cell death gene ced-3 whose overexpression can induce apoptosis in fibroblasts.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CASP-1; CASP1 nirs variant 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase-1; Caspase1; ICE; IL-1 beta-converting enzyme; IL-1BC; IL1B-convertase; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin-1 beta convertase; interleukin-1 beta converting enzyme; Interleukin-1 beta-converting enzyme; p45

View more View less

Gene Aliases: CASP1; ICE; IL1BC; IL1BCE; P45

View more View less

UniProt ID: (Human) P29466, (Pig) Q9N2I1

View more View less

Entrez Gene ID: (Human) 834, (Pig) 397319

View more View less

Function(s)
endopeptidase activity cysteine-type endopeptidase activity protein binding cysteine-type endopeptidase activator activity involved in apoptotic process cysteine-type endopeptidase activity involved in apoptotic process
Process(es)
response to hypoxia proteolysis apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process signal transduction regulation of autophagy protein processing response to lipopolysaccharide interleukin-1 beta production response to ATP regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of interleukin-1 alpha secretion positive regulation of interleukin-1 beta secretion regulation of inflammatory response mitochondrial depolarization membrane hyperpolarization pyroptosis cellular response to mechanical stimulus cellular response to organic substance programmed necrotic cell death toxin transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0